Skip to Content
Merck
All Photos(1)

Key Documents

63102

Sigma-Aldrich

Magnesium perchlorate

puriss., free-flowing powder, ≥99.0% (calc. based on dry substance, KT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Mg(ClO4)2
CAS Number:
Molecular Weight:
223.21
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
form:
free flowing powder

grade

puriss.

Assay

≥99.0% (calc. based on dry substance, KT)

form

free flowing powder

quality

free-flowing powder

impurities

≤8% water

storage temp.

room temp

SMILES string

[Mg++].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O

InChI

1S/2ClHO4.Mg/c2*2-1(3,4)5;/h2*(H,2,3,4,5);/q;;+2/p-2

InChI key

MPCRDALPQLDDFX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Magnesium perchlorate (Mg(ClO4)2) is widely used as drying agent for gases. It can remove water from gases (with no organic contaminants) at the rate of 0.001mgwater/l.

Application

Magnesium perchlorate (Mg(ClO4)2) may be employed as a catalyst in the following studies:
  • Preparation of a-aminophosphonates.
  • Enantioselective Diels-Alder reaction between cyclopentadiene and 3-acryloyl-1,3-oxazolin-2-one.
  • Preparation of imines and phenylhydrazones.
  • Protection of alcohols in the form of t-butyl ethers.
filler for drying tubes (especially for elemental analysis)

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magnesium perchlorate as an efficient catalyst for the synthesis of imines and phenylhydrazones.
Chakraborti AK, et al.
Tetrahedron Letters, 45(41), 7641-7644 (2004)
Eagleson M.
Concise Encyclopedia Chemistry, 343-343 (1994)
Giuseppe Bartoli et al.
Organic letters, 7(3), 427-430 (2005-01-28)
[reaction: see text] A new mild method for protecting alcohols as t-butyl ethers is reported. The reaction proceeds with Mg(ClO4)2 and Boc2O and shows general applicability. The deprotection of t-butyl ethers has also been revisited. Preliminary results indicate the CeCl3
The first enantioselective synthesis of both Diels-Alder enantiomers with the same bis (oxazoline)-magnesium perchlorate chiral catalyst.
Desimoni G, et al.
Tetrahedron Letters, 37(17), 3027-3030 (1996)
F Javier Céspedes-Guirao et al.
The Journal of organic chemistry, 74(16), 5871-5880 (2009-07-29)
Zinc phthalocyanine-perylenebisimide pentameric arrays, ZnPc(PDI)(4) 1 and 2, have been synthesized. ZnPc(PDI)(4) 1 has no substituents in the PDI bay positions, while ZnPc(PDI)(4) 2 presents four phenoxy groups in the bay positions of each perylene. In both cases, the PDI

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service