Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Magnesium perchlorate
Linear Formula:
Mg(ClO
4
)
2
CAS Number:
10034-81-8
Molecular Weight:
223.21
EC Number:
233-108-3
Products
Product Category
salts (3)
Brand
Sigma-Aldrich (3)
Formula Weight
Physical Form
Purity
Quality Segment
Reagent Type
Special Grade
ACS reagent (2)
Puriss (1)
Puriss P.A. (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
222283
Magnesium perchlorate,
ACS reagent
Expand
View Pricing
63095
Magnesium perchlorate,
puriss. p.a., drying agent, ACS reagent, ≥98.0% (calc. based on dry substance, KT)
Expand
View Pricing
63102
Magnesium perchlorate,
puriss., free-flowing powder, ≥99.0% (calc. based on dry substance, KT)
Expand
View Pricing