B21805
Dibenzyl disulfide
98%
Synonym(s):
Benzyl disulfide, Bis(phenylmethyl) disulfide
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
C6H5CH2SSCH2C6H5
CAS Number:
Molecular Weight:
246.39
Beilstein:
1110443
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
crystals
mp
69-72 °C (lit.)
SMILES string
C(SSCc1ccccc1)c2ccccc2
InChI
1S/C14H14S2/c1-3-7-13(8-4-1)11-15-16-12-14-9-5-2-6-10-14/h1-10H,11-12H2
InChI key
GVPWHKZIJBODOX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
R Munday et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 41(11), 1609-1615 (2003-09-10)
Thiols and disulphides make an important contribution to the organoleptic characteristics of foodstuffs, and many such compounds are approved for use as food flavours. Some thiols and disulphides are haemolytic agents in vivo. The haemolysis is caused by oxidative damage
Homan Kang et al.
Scientific reports, 5, 10144-10144 (2015-05-29)
Recently, preparation and screening of compound libraries remain one of the most challenging tasks in drug discovery, biomarker detection, and biomolecular profiling processes. So far, several distinct encoding/decoding methods such as chemical encoding, graphical encoding, and optical encoding have been
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service