Skip to Content
Merck
All Photos(4)

Documents

255823

Sigma-Aldrich

Lithium carbonate

ACS reagent, ≥99.0%

Synonym(s):

Carbolithium, Carbonic acid lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li2CO3
CAS Number:
Molecular Weight:
73.89
Beilstein:
3999191
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

Assay

≥99.0%

form

powder

impurities

≤0.01% insol. dil. HCl
≤0.2% S compounds

pH

9-11 (1 g/L)

mp

618 °C (lit.)

anion traces

chloride (Cl-): ≤0.005%
nitrate (NO3-): ≤5 ppm

cation traces

Ca: ≤0.01%
Fe: ≤0.002%
K: ≤0.01%
NH4+: ≤5 ppm
Na: ≤0.1%
heavy metals: ≤0.002% (by ICP-OES)

SMILES string

[Li+].[Li+].[O-]C([O-])=O

InChI

1S/CH2O3.2Li/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

XGZVUEUWXADBQD-UHFFFAOYSA-L

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Lithium carbonate is an inorganic compound and serves as the precursor for the synthesis of several lithium compounds including Li-ion battery materials. It is also used in the glass and ceramic industry.
Lithium carbonate is used
  • In the synthesis of monoclinic phase lithium zirconate by heating with a mixture of zirconia and potassium carbonate at high temperature.
  • As a lithium precursor to synthesize solid-base catalyst [Li4SiO4] by simple solid-state reaction with rice husks. Li4SiO4 is used for biodiesel production.
  • In the synthesis of high-purity lithium hydroxide monohydrate by membrane electrolysis process using lithium sulfate as electrolyte.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Towards homonuclear J solid-state NMR correlation experiments for half-integer quadrupolar nuclei: experimental and simulated 11B MAS spin-echo dephasing and calculated 2JBB coupling constants for lithium diborate.
Barrow NS, et al.
Physical Chemistry Chemical Physics, 13(13), 5778-5789 (2011)
Vanadium modified LiFePO4 cathode for Li-ion batteries.
Hong J, et al
Electrochemical and Solid-State Letters, 12(2), A33-A38 (2009)
Xiaoyue Tan et al.
Journal of the American Society of Nephrology : JASN, 17(12), 3382-3393 (2006-11-04)
Deficiency in vitamin D and its active metabolites is a pathologic feature of chronic kidney diseases. Despite that tubular epithelial cells are the major sites of active vitamin D synthesis, little is known about the role of vitamin D in
Hart WA, et al.
The Chemistry of Lithium, Sodium, Potassium, Rubidium, Cesium and Francium, 355-355 (1973)
Andrew A Nierenberg et al.
The American journal of psychiatry, 170(1), 102-110 (2013-01-05)
Lithium salts, once the mainstay of therapy for bipolar disorder, have tolerability issues at a higher dosage that often limit adherence. The authors investigated the comparative effectiveness of more tolerable dosages of lithium as part of optimized personalized treatment (OPT).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service