Skip to Content
Merck
All Photos(2)

Key Documents

203629

Sigma-Aldrich

Lithium carbonate

99.997% trace metals basis

Synonym(s):

Carbolithium, Carbonic acid lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li2CO3
CAS Number:
Molecular Weight:
73.89
Beilstein:
3999191
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38020204
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.997% trace metals basis
form:
powder

Quality Level

Assay

99.997% trace metals basis

form

powder

impurities

≤35.0 ppm Trace Metal Analysis

mp

618 °C (lit.)

density

2.11 g/mL at 25 °C

application(s)

battery manufacturing

SMILES string

[Li+].[Li+].[O-]C([O-])=O

InChI

1S/CH2O3.2Li/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

XGZVUEUWXADBQD-UHFFFAOYSA-L

Gene Information

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K E Morrison et al.
The Lancet. Neurology, 12(4), 339-345 (2013-03-05)
Lithium has neuroprotective effects in cell and animal models of amyotrophic lateral sclerosis (ALS), and a small pilot study in patients with ALS showed a significant effect of lithium on survival. We aimed to assess whether lithium improves survival in
Robert L Findling et al.
Journal of child and adolescent psychopharmacology, 23(2), 80-90 (2013-03-21)
This study examined the long-term effectiveness of lithium for the treatment of pediatric bipolar disorder within the context of combination mood stabilizer therapy for refractory mania and pharmacological treatment of comorbid psychiatric conditions. Outpatients, ages 7-17 years, meeting American Psychiatric
Smaranda Leu-Semenescu et al.
Neurology, 85(19), 1655-1662 (2015-10-11)
To compare the benefits and risks of lithium therapy vs abstention/other treatments in Kleine-Levin syndrome (KLS). In a KLS cohort followed in a single center, 130 patients regularly took lithium carbonate (median dose 1,000 mg/day; n = 71; 40 children)
N Alessi et al.
Journal of the American Academy of Child and Adolescent Psychiatry, 33(3), 291-304 (1994-03-01)
The use of lithium to treat child and adolescent psychiatric disorders is becoming more common. Since the publication of the report of The Committee on Biological Aspects of Child Psychiatry of the American Academy of Child Psychiatry in 1978, a
Andrea Contestabile et al.
The Journal of clinical investigation, 123(1), 348-361 (2012-12-04)
Down syndrome (DS) patients exhibit abnormalities of hippocampal-dependent explicit memory, a feature that is replicated in relevant mouse models of the disease. Adult hippocampal neurogenesis, which is impaired in DS and other neuropsychiatric diseases, plays a key role in hippocampal

Articles

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service