Skip to Content
Merck
All Photos(1)

Key Documents

601470

Sigma-Aldrich

Lithium-7Li2 carbonate

≥99 atom % 7Li, 99% (CP)

Synonym(s):

Lithium carbonate-7Li2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
7Li2CO3
CAS Number:
Molecular Weight:
74.04
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

≥99 atom % 7Li

Quality Level

Assay

99% (CP)

mp

720 °C (lit.)

mass shift

depleted

SMILES string

[7Li+].[7Li+].[O-]C([O-])=O

InChI

1S/CH2O3.2Li/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2/i;2*1+0

InChI key

XGZVUEUWXADBQD-LDAMREJGSA-L

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yuhua Duan et al.
Physical chemistry chemical physics : PCCP, 17(35), 22543-22547 (2015-08-19)
By combining TGA and XRD measurements with theoretical calculations of the capture of CO2 by lithium-rich zirconates (Li8ZrO6 and Li6Zr2O7), it has been demonstrated that the primary regeneration product during absorption/desorption cycling is in the form of Li2ZrO3. During absorption/desorption
Smaranda Leu-Semenescu et al.
Neurology, 85(19), 1655-1662 (2015-10-11)
To compare the benefits and risks of lithium therapy vs abstention/other treatments in Kleine-Levin syndrome (KLS). In a KLS cohort followed in a single center, 130 patients regularly took lithium carbonate (median dose 1,000 mg/day; n = 71; 40 children)
Rania I Nadeem et al.
Neurochemical research, 40(9), 1810-1818 (2015-07-29)
Recent studies have demonstrated a scrutinized association of diabetes mellitus with depressive symptoms and major depression. Glycogen synthase kinase-3 (GSK-3) is a protein kinase enzyme constitutively active in non-stimulated cells and in multiple signalings. Independent lines of research provide a
Mehmet K Kaya et al.
Clinical & experimental ophthalmology, 43(5), 449-457 (2015-02-03)
The aim of this study was to compare the effects of lapatinib and trastuzumab on vascular endothelial growth factor on experimental corneal neovascularization. A total of 35 Wistar albino rats were randomly divided into five groups, each containing seven animals.
Peter Visser et al.
Faraday discussions, 180, 511-526 (2015-05-01)
Lithium carbonate and lithium oxalate were incorporated as leachable corrosion inhibitors in model organic coatings for the protection of AA2024-T3. The coated samples were artificially damaged with a scribe. It was found that the lithium-salts are able to leach from

Articles

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Experts discuss challenges and production processes of nickel-rich layered oxide cathode materials in energy storage systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service