Skip to Content
Merck
All Photos(1)

Key Documents

Y0001361

Amphotericin B for microbiological assay

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Amphotericin B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H73NO17
CAS Number:
Molecular Weight:
924.08
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

amphotericin b

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

N[C@@H]1[C@@H]([C@@H](O[C@@H]([C@H]1O)C)O[C@@H]2C[C@@H]3O[C@@](C[C@@H]([C@H]3C(=O)O)O)(C[C@H](C[C@H]([C@@H](CC[C@H](C[C@H](CC(=O)O[C@H]([C@@H]([C@@H]([C@H](\C=C/C=C\C=C/C=C\C=C/C=C\C=C/2)C)O)C)C)O)O)O)O)O)O)O

InChI

1S/C47H73NO17/c1-27-17-15-13-11-9-7-5-6-8-10-12-14-16-18-34(64-46-44(58)41(48)43(57)30(4)63-46)24-38-40(45(59)60)37(54)26-47(61,65-38)25-33(51)22-36(53)35(52)20-19-31(49)21-32(50)23-39(55)62-29(3)28(2)42(27)56/h5-18,27-38,40-44,46,49-54,56-58,61H,19-26,48H2,1-4H3,(H,59,60)/b6-5-,9-7-,10-8-,13-11-,14-12-,17-15-,18-16-/t27-,28-,29-,30+,31+,32+,33-,34-,35+,36+,37-,38-,40+,41-,42+,43+,44-,46-,47+/m0/s1

InChI key

APKFDSVGJQXUKY-ZNVUZQDLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Amphotericin B for microbiological assay EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Renátó Kovács et al.
International journal of molecular sciences, 22(2) (2021-01-21)
Candida auris is a potential multidrug-resistant pathogen able to persist on indwelling devices as a biofilm, which serve as a source of catheter-associated infections. Neosartorya fischeri antifungal protein 2 (NFAP2) is a cysteine-rich, cationic protein with potent anti-Candida activity. We
Abeer H A Mohamed-Ahmed et al.
Current opinion in infectious diseases, 25(6), 695-702 (2012-11-14)
Amphotericin B (AmpB) is considered the first-line treatment for visceral leishmaniasis in areas in which resistance to antimony is prevalent. This review describes recent advances in clinically available and novel drug delivery systems of AmpB to treat visceral leishmaniasis. Over
Chen Nadler et al.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons, 72(5), 927-934 (2014-02-01)
The worldwide prevalence of leishmaniasis is increasing because of ecologic changes and increased medical profession awareness. Furthermore, solitary cases have been recently reported in Western countries. The authors describe the epidemiology, mode of transmission, and diagnosis of leishmaniasis and present
Manica Balasegaram et al.
Expert opinion on emerging drugs, 17(4), 493-510 (2012-11-22)
Leishmaniasis is a parasitic disease transmitted by phlebotomine sandflies. Between 700,000 and 1.2 million cases of cutaneous leishmaniasis and between 200,000 and 400,000 cases of visceral leishmaniasis (VL), which is fatal if left untreated, occur annually worldwide. Liposomal amphotericin B
Amphotericin B still in the headlines.
Luigina Romani
Pathogens and global health, 106(2), 80-81 (2012-09-05)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service