Skip to Content
Merck
All Photos(1)

Documents

SML2288

Sigma-Aldrich

Anidulafungin

≥97% (HPLC), powder, antifungal

Synonym(s):

1-[(4R,5R)-4,5-Dihydroxy-N2-[[4′′-(pentyloxy)[1,1′:4′,1′′-terphenyl]-4-yl]carbonyl]-L-ornithine]echinocandin B, LY 303366

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C58H73N7O17
CAS Number:
Molecular Weight:
1140.24
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

product name

Anidulafungin, ≥97% (HPLC)

Assay

≥97% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

CCCCCOC1=CC=C(C2=CC=C(C3=CC=C(C(N[C@H]4C[C@@H](O)[C@@H](O)NC([C@@H]5[C@@H](O)[C@@H](C)CN5C([C@H]([C@H](O)C)NC([C@H]([C@H](O)[C@@H](O)C6=CC=C(O)C=C6)NC([C@@H]7C[C@@H](O)CN7C([C@H]([C@H](O)C)NC4=O)=O)=O)=O)=O)=O)=O)C=C3)C=C2)C=C1

InChI

1S/C58H73N7O17/c1-5-6-7-24-82-40-22-18-35(19-23-40)33-10-8-32(9-11-33)34-12-14-37(15-13-34)51(74)59-41-26-43(70)54(77)63-56(79)47-48(71)29(2)27-65(47)58(81)45(31(4)67)61-55(78)46(50(73)49(72)36-16-20-38(68)21-17-36)62-53(76)42-25-39(69)28-64(42)57(80)44(30(3)66)60-52(41)75/h8-23,29-31,39,41-50,54,66-73,77H,5-7,24-28H2,1-4H3,(H,59,74)(H,60,75)(H,61,78)(H,62,76)(H,63,79)/t29-,30+,31+,39+,41-,42-,43+,44-,45-,46-,47-,48-,49-,50-,54+/m0/s1

InChI key

JHVAMHSQVVQIOT-MFAJLEFUSA-N

Application

Anidulafungin has been used as an internal positive inhibitor control in antifungal minimum inhibitory concentration (MIC) assay. It has also been used as an antifungal drug in disk diffusion assay to study drug resistance and tolerance in Candida spp.

Biochem/physiol Actions

Anidulafungin contains an alkoxytriphenyl chain, which may contribute to its anti-fungal activity. Anidulafungin may exhibit therapeutic effect against oesophageal and invasive candidiasis.
Anidulafungin is a semisynthetic echinocandin antifungal. Anidulafungin works by inhibiting the enzyme β(1,3)-D-Glucan synthase and thereby disturbing the integrity of the fungal cell wall. This enzyme does not exist in mammalian systems.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2

Target Organs

Liver

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Orawan Tulyaprawat et al.
Frontiers in microbiology, 11, 934-934 (2020-06-09)
Candidemia, a bloodstream infection caused by genus Candida, has a high mortality rate. Candida albicans was previously reported to be the most common causative species among candidemia patients. However, during the past 10 years in Thailand, Candida tropicalis has been
Youcef Megri et al.
Antimicrobial resistance and infection control, 9(1), 50-50 (2020-04-09)
Despite being associated with a high mortality and economic burden, data regarding candidemia are scant in Algeria. The aim of this study was to unveil the epidemiology of candidemia in Algeria, evaluate the antifungal susceptibility pattern of causative agents and
Ali Ahmadi et al.
Journal of global antimicrobial resistance, 21, 331-334 (2019-11-13)
Candida parapsilosis (C. parapsilosis) is a common non-albicans Candida species ranked as the second common cause of bloodstream infections. Azole resistance and elevated echinocandin MICs have been reported for these fungi. This study was conducted to determine the interactions between
Hossein Mirhendi et al.
Medical mycology, 58(2), 201-206 (2019-05-22)
As data on pediatric invasive candidiasis (IC) and the antifungal susceptibility pattern of associated isolates are scarce in Iran, this study aimed to determine species distribution and antifungal susceptibility profile of Candida species isolated from pediatric patients with suspected or
Ana Emília M Roberto et al.
Microorganisms, 8(1) (2020-01-17)
Mass spectrometry by matrix-assisted laser desorption/ionization time-of-flight (MALDI-TOF) was used to identify and differentiate the pattern of susceptibility of clinical isolates of Candida parapsilosis complex. 17 C. parapsilosis sensu stricto, 2 C. orthopsilosis, and 1 C. metapsilosis strains were obtained

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service