Skip to Content
Merck
All Photos(1)

Key Documents

Y0001014

Amphotericin B for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Amphotericin B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H73NO17
CAS Number:
Molecular Weight:
924.08
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

amphotericin b

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

N[C@@H]1[C@@H]([C@@H](O[C@@H]([C@H]1O)C)O[C@@H]2C[C@@H]3O[C@@](C[C@@H]([C@H]3C(=O)O)O)(C[C@H](C[C@H]([C@@H](CC[C@H](C[C@H](CC(=O)O[C@H]([C@@H]([C@@H]([C@H](\C=C/C=C\C=C/C=C\C=C/C=C\C=C/2)C)O)C)C)O)O)O)O)O)O)O

InChI

1S/C47H73NO17/c1-27-17-15-13-11-9-7-5-6-8-10-12-14-16-18-34(64-46-44(58)41(48)43(57)30(4)63-46)24-38-40(45(59)60)37(54)26-47(61,65-38)25-33(51)22-36(53)35(52)20-19-31(49)21-32(50)23-39(55)62-29(3)28(2)42(27)56/h5-18,27-38,40-44,46,49-54,56-58,61H,19-26,48H2,1-4H3,(H,59,60)/b6-5-,9-7-,10-8-,13-11-,14-12-,17-15-,18-16-/t27-,28-,29-,30+,31+,32+,33-,34-,35+,36+,37-,38-,40+,41-,42+,43+,44-,46-,47+/m0/s1

InChI key

APKFDSVGJQXUKY-ZNVUZQDLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Amphotericin B for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gurpreet Singh et al.
Journal of pediatric hematology/oncology, 35(3), e127-e129 (2013-03-21)
Hemophagocytic lymphohistiocytosis (HLH) associated with visceral leishmaniasis (VL) is a very rare phenomenon. We report the first known North American case in a 21 month old boy. He was initially diagnosed with Epstein Barr virus (EBV) triggered HLH and treated
Benoit Pilmis et al.
Medecine sciences : M/S, 29 Spec No 1, 25-30 (2013-05-15)
Mucormycosis is a life-threatening invasive fungal infection that arises among immunocompromised patients (haematological malignancies, solid organ transplantation, diabetes mellitus). The most frequent sites of infection are pulmonary, rhinocerebral, cutaneous and disseminated. Reversal of the underlying conditions is mandatory for controlling
Anil Bhansali et al.
The Journal of the Association of Physicians of India, 60, 54-58 (2013-06-20)
Histoplasmosis has been sporadically reported from India. Though asymptomatic adrenal involvement has been described in patients with disseminated histoplasmosis; isolated adrenal involvement with adrenal insufficiency (AI) as the presenting manifestation of the disease is rare. We describe 5 immunocompetent men
Afaq Ahmad Khan et al.
Journal of the Indian Medical Association, 110(7), 499-500 (2013-03-26)
Non-Hodgkin's lymphoma (NHL) is predominantly a disease of lymph nodes, but extranodal involvement is not very uncommon. Palatal involvement by NHL is rare. Mucormycosis is a devastating fungal infection commonly seen in immunocompromised individuals, including those with NHL, but it
John E Linley
Methods in molecular biology (Clifton, N.J.), 998, 149-157 (2013-03-27)
Perforated whole-cell patch-clamp is a variant of the patch-clamp technique used to measure the sum activity of ion channels in the plasma membrane of a single cell. Its defining feature is that electrical access to the cell is obtained through

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service