Skip to Content
Merck
All Photos(1)

Key Documents

S6814

Sigma-Aldrich

SynaptoGreen C4

≥95% purity (HPLC), solid

Synonym(s):

FM1-43, N-(3-triethylammoniumpropyl) -4-(4-(dibutylamino)styryl) pyridinium dibromide, Pyridinium, 4-[2-[4-(dibutylamino)phenyl]ethenyl]- 1-[3-(triethylammonio)propyl]- , dibromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H49Br2N3
CAS Number:
Molecular Weight:
611.54
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

SynaptoGreen C4, ≥95% (HPLC), solid

Quality Level

Assay

≥95% (HPLC)

form

solid

color

dark red

solubility

DMSO: soluble
H2O: soluble

εmax

≥50000 at 507-513 nm in methanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

−20°C

SMILES string

[Br-].[Br-].CCCCN(CCCC)c1ccc(\C=C\c2cc[n+](CCC[N+](CC)(CC)CC)cc2)cc1

InChI

1S/C30H49N3.2BrH/c1-6-11-23-32(24-12-7-2)30-18-16-28(17-19-30)14-15-29-20-25-31(26-21-29)22-13-27-33(8-3,9-4)10-5;;/h14-21,25-26H,6-13,22-24,27H2,1-5H3;2*1H/q+2;;/p-2

InChI key

VZUVCAGXYLMFEC-UHFFFAOYSA-L

General description

SynaptoGreen C4 is an oxonol dye that helps in the rapid detection of an antibiotic′s effect on bacterial viability. It is a fluorescent dye that is used in optical imaging studies to determine synaptic activity by staining synaptic vesicles at the nerve terminal or the neuromuscular junction. It is equivalent to FM1-43.

Application

SynaptoGreen C4 has been used to label cultured neurons grown on EM grids for fluorescent labeling, live light microscopy and imaging studies.

Biochem/physiol Actions

A fluorescent nerve terminal dye suitable for monitoring synaptic activity at synapses and at neuromuscular junctions.

Other Notes

Component of NTSI, also sold individually.

Legal Information

SynaptoGreen is a trademark of Biotium, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roman Pleskot et al.
Frontiers in plant science, 3, 54-54 (2012-05-29)
Phosphatidic acid (PA) is an important intermediate in membrane lipid metabolism that acts as a key component of signaling networks, regulating the spatio-temporal dynamics of the endomembrane system and the cytoskeleton. Using tobacco pollen tubes as a model, we addressed
Journal of Immunology, 179(2) null
Christopher R Heier et al.
EMBO molecular medicine, 5(10), 1569-1585 (2013-09-10)
Absence of dystrophin makes skeletal muscle more susceptible to injury, resulting in breaches of the plasma membrane and chronic inflammation in Duchenne muscular dystrophy (DMD). Current management by glucocorticoids has unclear molecular benefits and harsh side effects. It is uncertain
Yuniar Devi Utami et al.
The ISME journal, 13(2), 455-467 (2018-10-06)
We investigated the phylogenetic diversity, localisation and metabolism of an uncultured bacterial clade, Termite Group 2 (TG2), or ZB3, in the termite gut, which belongs to the candidate phylum 'Margulisbacteria'. We performed 16S rRNA amplicon sequencing analysis and detected TG2/ZB3
Santiago Balseiro-Gomez et al.
Journal of cell science, 129(21), 3989-4000 (2016-11-03)
To ensure normal immune function, mast cells employ different pathways to release mediators. Here, we report a thus far unknown capacity of mast cells to recycle and reuse secretory granules after an antigen-evoked degranulation process under physiological conditions; this phenomenon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service