Skip to Content
Merck
All Photos(2)

Documents

F4018

Sigma-Aldrich

Anti-Mouse IgG (Fab specific)−FITC antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Mouse IgG (Fab specific)−Fluorescein isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

FITC conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

storage condition

protect from light

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:200
indirect immunofluorescence: 1:64

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgG (immunoglobulin G) is found in four subclasses IgG1, IgG2, IgG3 and IgG4. IgG has a monomeric structure.
IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections.

Specificity

Anti-Mouse IgG (Fab specific)-FITC antibody is specific for Fab fragment of all mouse IgG subclasses. Goat anti-Mouse IgG is conjugated to FITC and then purified by gel filtration to remove free FITC. The antibody is adsorbed with human IgG to ensure minimal cross reactivity. The conjugate shows no reactivity with mouse IgG, Fc fragment, human, bovine, or horse serum.

Immunogen

Purified mouse IgG Fab fragment

Application

Anti-Mouse IgG (Fab specific) FITC antibody produced in goat has been used in :
  • fluorescent staining
  • immunocytochemistry
  • immunofuorescence

Biochem/physiol Actions

IgG (immunoglobulin G) antibody provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections. IgG antibody participates in complement fixation and opsonization.

Other Notes

Antibody adsorbed with bovine, equine and human serum proteins.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Preparation Note

Adsorbed to reduce background staining with bovine, horse, or human samples.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Changes in erythropoiesis in hereditary hemochromatosis are not mediated by HFE expression in nucleated red cells
Feeney GP, et al.
Haematologica, 90(2), 180-187 (2005)
Amino-terminally truncated desmin rescues fusion of des-/- myoblasts but negatively affects cardiomyogenesis and smooth muscle development
Hollrigl A, et al.
Febs Letters, 523(1-3), 229-233 (2002)
Susann Erdmann et al.
Eukaryotic cell, 11(5), 571-589 (2012-01-03)
In this study, we undertook a functional characterization and transcriptome analysis that enabled a comprehensive study of the mating type loci of the mushroom Schizophyllum commune. Induced expression of both the bar2 receptor and the bap2(2) pheromone gene within 6
Complex regulation of spiral ganglion neuron firing patterns by neurotrophin-3
Zhou Z, et al.
The Journal of Neuroscience, 25(33), 7558-7566 (2005)
Xiuli Wang et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 24(1), 106-119 (2017-10-25)
Purpose: Multiple myeloma remains an incurable malignancy of plasma cells despite considerable advances in treatment. The purpose of the study was to develop novel chimeric antigen receptors (CAR) for the treatment of multiple myeloma and explore combinatorial therapy using CAR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service