Skip to Content
Merck
All Photos(1)

Key Documents

PHR1088

Supelco

Azithromycin

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Azithromycin dihydrate, 1-Oxa-6-azacyclopentadecan-15-one, 13-[(2,6-dideoxy-3-C-methyl-3-O-methyl-alpha-L-ribo-hexopyranosyl)oxy]-2-ethyl-3,4,10- trihydroxy-3,5,6,8,10,12,14-heptamethyl-11-[[3,4,6-trideoxy-3-(dimethylamino)-beta-D-xylohexopyranosyl]oxy]-, [2R-(2R, 3S, 4R, 5R, 8R, 10R, 11R, 12S, 13S, 14R)]- dihydrate, N-Methyl-11-aza-10-deoxo-10-dihydroerythromycin A, CP-62993

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H72N2O12 · 2H2O
CAS Number:
Molecular Weight:
785.02
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0000306
traceable to USP 1046056

API family

azithromycin

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

-10 to -25°C

SMILES string

O.O.CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@](C)(O)C[C@@H](C)CN(C)[C@H](C)[C@@H](O)[C@]1(C)O

InChI

1S/C38H72N2O12.2H2O/c1-15-27-38(10,46)31(42)24(6)40(13)19-20(2)17-36(8,45)33(52-35-29(41)26(39(11)12)16-21(3)48-35)22(4)30(23(5)34(44)50-27)51-28-18-37(9,47-14)32(43)25(7)49-28;;/h20-33,35,41-43,45-46H,15-19H2,1-14H3;2*1H2/t20-,21-,22+,23-,24-,25+,26+,27-,28+,29-,30+,31-,32+,33-,35+,36-,37-,38-;;/m1../s1

InChI key

SRMPHJKQVUDLQE-KUJJYQHYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Azithromycin may be used as a pharmaceutical reference standard for the determination of the analyte in plasma/serum samples and pharmaceutical formulations by various chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Azithromycin dihydrate is a macrolide antibiotic with anti-immunomodulatory/anti-inflammatory properties.
Azithromycin dihydrate is a macrolide antibiotic, azalide subclass. It binds to the 50S subunit of the 70S bacterial ribosomes and inhibits RNA-dependent protein synthesis in bacterial cells. Azithromycin also has anti-immunomodulatory/anti-inflammatory properties, which make it useful in treating cystic fibrosis.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB3758 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative determination of the macrolide antibiotics erythromycin, roxithromycin, azithromycin and clarithromycin in human serum by high-performance liquid chromatography using pre-column derivatization with 9-fluorenylmethyloxycarbonyl chloride and fluorescence detection
Torano JS and Guchelaar HJ
Journal of Chromatography. B, Biomedical Sciences and Applications, 720(1-2), 89-97 (1998)
Determination of erythromycin, clarithromycin, roxithromycin, and azithromycin in plasma by high-performance liquid chromatography with amperometric detection
Taninaka C, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 738(2), 405-411 (2000)
LC determination of impurities in azithromycin tablets
Miguel L and Barbas C
Journal of Pharmaceutical and Biomedical Analysis, 33(2), 211-217 (2003)
Robin Vos et al.
Transplantation, 94(2), 101-109 (2012-03-31)
Chronic lung allograft rejection is the single most important cause of death in lung transplant recipients after the first postoperative year, resulting in a 5-year survival rate of approximately 50%, which is far behind that of other solid organ transplantations.
Erik B Wilms et al.
Pediatric pulmonology, 47(7), 658-665 (2012-06-12)
Azithromycin maintenance therapy results in improvement of respiratory function in patients with cystic fibrosis (CF). In azithromycin maintenance therapy, several dosing schemes are applied. In this review, we combine current knowledge about azithromycin pharmacokinetics with the dosing schedules used in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service