Skip to Content
Merck
All Photos(1)

Key Documents

1046078

USP

Desosaminylazithromycin

United States Pharmacopeia (USP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H58N2O9
CAS Number:
Molecular Weight:
590.79
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

azithromycin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

N1([C@@H]([C@H]([C@@]([C@H](OC(=O)[C@@H]([C@H]([C@@H]([C@H]([C@@](C[C@H](C1)C)(O)C)O[C@@H]2O[C@@H](C[C@@H]([C@H]2O)N(C)C)C)C)O)C)CC)(O)C)O)C)C

InChI

1S/C30H58N2O9/c1-12-22-30(8,38)25(35)20(6)32(11)15-16(2)14-29(7,37)26(18(4)23(33)19(5)27(36)40-22)41-28-24(34)21(31(9)10)13-17(3)39-28/h16-26,28,33-35,37-38H,12-15H2,1-11H3/t16-,17-,18+,19-,20-,21+,22-,23+,24-,25-,26-,28+,29-,30-/m1/s1

InChI key

PXDYILJJHOVNLO-NZMWSZMZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Desosaminylazithromycin USP reference standard intended for use in specified quality tests and assays.

Also used to prepare standard stock, system suitability, standard, system suitability stock, and peak identification solution for impurity analysis according to the given below monographs of United States Pharmacopeia (USP):
  • Azithromycin for Injection [1]
  • Azithromycin [2]
  • Azithromycin Tablets [3]
  • Azithromycin for Oral Suspension [4]

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Azithromycin for Injection
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 44(2), 451-451 (2020)
Azithromycin
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 43(3), 446-446 (2020)
Azithromycin Tablets
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 44(3), 457-457 (2020)
Azithromycin for Oral Suspension
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 44(5) (2022)
Fuad Al-Rimawi et al.
Journal of chromatographic science, 48(2), 86-90 (2010-01-30)
A simple, validated stability-indicating liquid chromatographic method is developed for the analysis of azithromycin in raw material and in pharmaceutical forms. Liquid chromatography with a UV detector at a wavelength of 210 nm using a reversed-phase C(18) stationary phase has

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service