Skip to Content
Merck
All Photos(1)

Key Documents

A3487

Sigma-Aldrich

Clarithromycin

96.0-102.0% (HPLC)

Synonym(s):

6-O-Methylerythromycin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H69NO13
CAS Number:
Molecular Weight:
747.95
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85

biological source

synthetic

Quality Level

Assay

96.0-102.0% (HPLC)

form

powder

color

white to off-white

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

O[C@@H]([C@@](O)(C)[C@@H](CC)OC([C@H](C)[C@H]1O[C@@H]2O[C@@H](C)[C@H](O)[C@](C)(OC)C2)=O)[C@@H](C)C([C@H](C)C[C@@](C)(OC)[C@H](O[C@@H]3O[C@H](C)C[C@H](N(C)C)[C@H]3O)[C@H]1C)=O

InChI

1S/C38H69NO13/c1-15-26-38(10,45)31(42)21(4)28(40)19(2)17-37(9,47-14)33(52-35-29(41)25(39(11)12)16-20(3)48-35)22(5)30(23(6)34(44)50-26)51-27-18-36(8,46-13)32(43)24(7)49-27/h19-27,29-33,35,41-43,45H,15-18H2,1-14H3/t19-,20-,21+,22+,23-,24+,25+,26-,27+,29-,30+,31-,32+,33-,35+,36-,37-,38-/m1/s1

InChI key

AGOYDEPGAOXOCK-KCBOHYOISA-N

Looking for similar products? Visit Product Comparison Guide

Application

Clarithromycin has been used to study its effect on the potassium channel KCNJ5WT.

Biochem/physiol Actions

Clarithromycin is a macrolide antibiotic that interferes with bacterial protein synthesis. Clarithromycin is an acid-stable version of erythromycin and is particularly effective against Gram-negative bacteria. It has a short half-life, however its metabolite, 14-hydroxy clarithromycin is nearly twice as active as clarithromycin against certain bacteria.
Clarithromycin is stable in gastric acid.
Clarithromycin possess bactericidal activity including anti-mycobacterial activity against Mycobacterium leprae, and M.avium complex.

Packaging

100mg

Preparation Note

Practically insoluble in water, soluble in acetone and methylene chloride, slightly soluble in methanol.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rui Chen et al.
Drug delivery, 24(1), 1191-1203 (2017-08-29)
Inhalation administration, compared with intravenous administration, significantly enhances chemotherapeutic drug exposure to the lung tissue and may increase the therapeutic effect for pulmonary anticancer. However, further identification of cancer cells after lung deposition of inhaled drugs is necessary to avoid
Leprosy
Antibiotic and chemotherapy e-book (2010)
Macrolides selectively inhibit mutant KCNJ5 potassium channels that cause aldosterone-producing adenoma
Scholl UI, et al.
The Journal of Clinical Investigation, 127(7), 2739-2750 (2017)
Sonia Agudo et al.
Journal of clinical microbiology, 48(10), 3703-3707 (2010-07-30)
Clarithromycin is one of the antibiotics used for the treatment of Helicobacter pylori infections, and clarithromycin resistance is the most important factor when it comes to predicting eradication failure. The present study analyzed H. pylori isolates for the presence of
Antituberculosis drugs
Antibiotic and chemotherapy e-book (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service