Skip to Content
Merck
All Photos(1)

Key Documents

I1892

Supelco

Ibuprofen sodium salt

≥98% (GC)

Synonym(s):

α-Methyl-4-(isobutyl)phenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C13H17O2Na
CAS Number:
Molecular Weight:
228.26
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98% (GC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

solubility

H2O: 100 mg/mL, clear to slightly hazy

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

[Na+].CC(C)Cc1ccc(cc1)C(C)C([O-])=O

InChI

1S/C13H18O2.Na/c1-9(2)8-11-4-6-12(7-5-11)10(3)13(14)15;/h4-7,9-10H,8H2,1-3H3,(H,14,15);/q;+1/p-1

InChI key

PTTPUWGBPLLBKW-UHFFFAOYSA-M

Gene Information

General description

Ibuprofen belongs to the class of non-steroidal anti-inflammatory drugs, widely used for the treatment of mild to moderate pain and several forms of arthritis.

Application

Ibuprofen sodium salt may be used as a model drug to study the intercalation mechanism via ion exchange reaction between ibuprofen and a biocompatible hydrotalcite compound.
Ibuprofen sodium salt may be used as an analytical reference standard for the qualification of the analyte in wastewaters using high-performance liquid chromatography (HPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Cyclooxygenase (COX) inhibitor that has greater activity against COX-1 than against COX-2.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fareeha Batool et al.
Materials (Basel, Switzerland), 11(4) (2018-04-13)
Ibuprofen (IBU) has been shown to improve periodontal treatment outcomes. The aim of this study was to develop a new anti-inflammatory scaffold by functionalizing an electrospun nanofibrous poly-ε-caprolactone membrane with IBU (IBU-PCL) and to evaluate its impact on periodontal inflammation
Mixed supramolecular hemimicelles aggregates and magnetic carrier technology for solid phase extraction of ibuprofen in environmental samples prior to its HPLC-UV determination
Beiraghi A, et al.
Chemical Engineering Science, 108(3), 103-110 (2014)
David J Pérez et al.
European journal of pharmacology, 814, 18-27 (2017-08-02)
There are many chronic diseases related with inflammation. The chronic inflammation can produce other problems as cancer. Therefore, it is necessary to design drugs with better anti-inflammatory activity than those in the clinic. Likewise, these could be used in chronic
Abegaz Tizazu Andrgie et al.
Polymers, 12(11) (2020-11-12)
Hydrogels have been investigated as ideal biomaterials for wound treatment owing to their ability to form a highly moist environment which accelerates cell migration and tissue regeneration for prompt wound healing. They can also be used as a drug carrier
Determination of non-steroidal anti-inflammatory drugs in wastewaters by magnetic matrix solid phase dispersion?HPLC
Aguilar-Arteaga.K, et al.
Talanta, 80(3), 1152-1157 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service