Skip to Content
Merck
All Photos(1)

Key Documents

PHR1435

Supelco

Warfarin Sodium

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

3-(α-Acetonylbenzyl)-4-hydroxycoumarin sodium salt, 4-Hydroxy-3-(3-oxo-1-phenylbutyl)coumarin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C19H15O4Na
CAS Number:
Molecular Weight:
330.31
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 672
traceable to Ph. Eur. W0100000
traceable to USP 1719000

API family

warfarin

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na].CC(=O)CC(c1ccccc1)C2=C(O)c3ccccc3OC2=O

InChI

1S/C19H16O4.Na/c1-12(20)11-15(13-7-3-2-4-8-13)17-18(21)14-9-5-6-10-16(14)23-19(17)22;/h2-10,15,21H,11H2,1H3;/q;+1/p-1

InChI key

KYITYFHKDODNCQ-UHFFFAOYSA-M

Gene Information

human ... VKORC1(79001)

Looking for similar products? Visit Product Comparison Guide

General description

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to in-house working standards. Warfarin Sodium is an anticoagulant rodenticide, commonly used for the control of rodents such as rats, mice, etc. in surroundings, commercial and industrial sites.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.
Warfarin Sodium may be used pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using square-wave adsorptive cathodic stripping voltammetry.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA2743 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Chronic 3 - Repr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lippincott's Review for NCLEX-RN, 511(1), 63-69 (2005)
Analytical Methods for Therapeutic Drug Monitoring and Toxicology (2011)
Saunders Handbook of Veterinary Drugs: Small and Large Animal (2015)
Colby A Nadeau et al.
Translational stroke research, 10(3), 287-297 (2018-06-28)
Intracerebral hemorrhage (ICH) causes blood-brain barrier (BBB) damage along with altered element levels in the brain. BBB permeability was quantified at 3, 7, and 14 days with Evans Blue dye after collagenase-induced ICH in rat. At peak permeability (day 3), a
Urszula Śliwińska-Hill et al.
Journal of biomolecular structure & dynamics, 37(14), 3731-3739 (2018-09-22)
The interaction between glycated human serum albumin (gHSA) and folic acid (FA) was investigated by various spectroscopic techniques, such as fluorescence, circular dichroism, UV-vis absorption spectroscopy and electrophoretic light scattering technique. These methods characterize the binding properties of an albumin-folic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service