Skip to Content
Merck
All Photos(4)

Key Documents

D4784

Sigma-Aldrich

4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride

≥98% purity (HPLC), powder

Synonym(s):

1,2-Diamino-4,5-methylenedioxybenzene dihydrochloride, 1,3-Benzodioxole-5,6-diamine dihydrochloride, 5,6-Diamino-1,3-benzodioxole dihydrochloride, MDB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8N2O2 · 2HCl
CAS Number:
Molecular Weight:
225.07
Beilstein:
5641607
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

Product Name

4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride, Fluorogenic reagent

Quality Level

Assay

≥98% (HPLC)

form

powder

technique(s)

titration: suitable

color

white to orange

mp

247 °C

solubility

H2O: 50 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

−20°C

SMILES string

Cl[H].Cl[H].Nc1cc2OCOc2cc1N

InChI

1S/C7H8N2O2.2ClH/c8-4-1-6-7(2-5(4)9)11-3-10-6;;/h1-2H,3,8-9H2;2*1H

InChI key

YXEJRYIEFFUUID-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride is a fluorescence tag for α keto acids. It is also required for the derivatization of sialic acids in presence of dilute sulfuric acid. The method allows determination of sialic acids.[1]

Application

4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride has been used for the derivatization of sialic acids to analyse them by capillary liquid-chromatography-electrospray ionization/tandem mass spectrometry (CapLC-ESI-MS/MS).[2][3]
Fluorogenic reagent for α-ketoacids.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Capillary Electrophoresis of Carbohydrates (2010)
Determination of the type and quantity of sialic acid in the egg jelly coat of the sea urchin Paracentrotus lividus using capillary LC-ESI-MS/MS.
Yesilyurt B, et al.
Molecular Reproduction and Development, 82, 115-122 (2015)
Determination of sialic acids in immune system cells (coelomocytes) of sea urchin, Paracentrotus lividus, using capillary LC-ESI-MS/MS.
Izzetoglu S, et al.
Fish & Shellfish Immunology, 36, 181-186 (2014)
Savaş İzzetoğlu et al.
Fish & shellfish immunology, 36(1), 181-186 (2013-11-13)
Coelomocytes are considered to be immune effectors of sea urchins. Coelomocytes are the freely circulating cells in the body fluid contained in echinoderm coelom and mediate the cellular defence responses to immune challenges by phagocytosis, encapsulation, cytotoxicity and the production
Xiaoxiao Guo et al.
The Analyst, 145(13), 4512-4521 (2020-05-16)
The polysialyltransferases (polySTs) catalyse the polymerisation of polysialic acid, which plays an important role in tumour metastasis. While assays are available to assess polyST enzyme activity, there is no methodology available specifically optimised for identification and quantitative evaluation of potential

Articles

A significantly improved HPLC-fluorescence method for DMB-NANA and -NGNA, and application of this method to compare 2 candidate biosimilar therapeutic proteins to their respective RMs.

A significantly improved HPLC-fluorescence method for DMB-NANA and -NGNA, and application of this method to compare 2 candidate biosimilar therapeutic proteins to their respective RMs.

A significantly improved HPLC-fluorescence method for DMB-NANA and -NGNA, and application of this method to compare 2 candidate biosimilar therapeutic proteins to their respective RMs.

A significantly improved HPLC-fluorescence method for DMB-NANA and -NGNA, and application of this method to compare 2 candidate biosimilar therapeutic proteins to their respective RMs.

Questions

  1. When running on HPLC after labeling, which option should be chosen between D4784 4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride and 66807 4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride?

    1 answer
    1. Both D4784 ,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride and 66807 4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride are not marketed as amino acid labeling products. D4784 has a purity of 98% as determined by HPLC, while 66807 has a purity of 97% according to HPLC analysis. Both products are essentially the same.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service