Skip to Content
Merck
All Photos(4)

Key Documents

157953

Sigma-Aldrich

Sodium hydrosulfite

technical grade

Synonym(s):

Sodium hypodisulfite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaO2SSO2Na
CAS Number:
Molecular Weight:
174.11
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥82.5% (iodometric)
grade:
technical grade
form:
powder
solubility:
water: soluble(lit.)

grade

technical grade

Quality Level

Assay

≥82.5% (iodometric)

form

powder

pH

7-9 (20 °C, 50 g/L)

solubility

water: soluble(lit.)

cation traces

heavy metals: ≤0.002%

storage temp.

room temp

SMILES string

[Na+].[Na+].[O-]S(=O)S([O-])=O

InChI

1S/2Na.H2O4S2/c;;1-5(2)6(3)4/h;;(H,1,2)(H,3,4)/q2*+1;/p-2

InChI key

JVBXVOWTABLYPX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Sodium hydrosulfite (Sodium dithionite, Sodium hypodisulfite, Na2S2O4) is a white crystalline sodium salt. It decomposes in the presence of atmospheric oxygen to afford sodium hydrogen sulfate. It has been prepared by reducing sodium hydrogen sulfite in sulfurous acid solution with zinc. It is widely used for the decolorization prior to dyeing of cloths in the textile industry.

Application

Sodium hydrosulfite may be employed as a reducing agent for the reduction of graphite oxide in a study.
Sodium hydrosulfite has been used as a fluorescent quenching agent in the encapsulation of polynucleotides via production of asymmetric nanoparticles. It has also be used in the form of buffer for the elution of probe labelled proteins involved in the indentification of cellular targets of covalent kinase inhibitors via mass spectrometry method (MS).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Self-heat. 1

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

E Gabriele Panarelli et al.
Physical chemistry chemical physics : PCCP, 22(17), 9487-9493 (2020-04-22)
We report on a novel approach to the study of rates and short-lived intermediates of (bio)chemical reactions that involve paramagnetic species. Temperature-cycle Electron Paramagnetic Resonance (EPR) concerns the repeated heating of a reaction mixture in the cavity of an EPR
A chemoproteomic method for identifying cellular targets of covalent kinase inhibitors
Chen C-Y and Zhang C
Genes & Cancer, 7, 148-148 (2016)
Evaluation of asymmetric liposomal nanoparticles for encapsulation of polynucleotides
Whittenton J, et al.
Langmuir, 24(16), 8533-8540 (2008)
The preparation of high performance and conductive poly (vinyl alcohol)/graphene nanocomposite via reducing graphite oxide with sodium hydrosulfite.
Zhou T, et al.
Composites Science and Technology, 71(9), 1266-1270 (2011)
A Siri Luthman et al.
Journal of biomedical optics, 24(3), 1-14 (2018-10-26)
Emerging clinical interest in combining standard white light endoscopy with targeted near-infrared (NIR) fluorescent contrast agents for improved early cancer detection has created demand for multimodal imaging endoscopes. We used two spectrally resolving detector arrays (SRDAs) to realize a bimodal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service