Skip to Content
Merck
All Photos(1)

Key Documents

BCR310

3-Nitrofluoranthene

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H9NO2
CAS Number:
Molecular Weight:
247.25
Beilstein:
2216474
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

157-159 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc2-c3ccccc3-c4cccc1c24

InChI

1S/C16H9NO2/c18-17(19)15-9-8-13-11-5-2-1-4-10(11)12-6-3-7-14(15)16(12)13/h1-9H

InChI key

PIHGQKMEAMSUNA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Nitrofluoranthene, belonging to the class of nitrated-polycyclic aromatic hydrocarbons, is persistent in the environment. It is produced from direct sources such as diesel, gasoline exhaust and gas-phase reactions of polycyclic aromatic hydrocarbons (PAHs) with oxides of nitrogen.

Analysis Note

For more information please see:
BCR310

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C E Mitchell et al.
Carcinogenesis, 14(6), 1161-1166 (1993-06-01)
The nitrofluoranthene (NF) family of compounds includes the potent pulmonary carcinogen 3,9-dinitrofluoranthene (3,9-DNF) and the weak carcinogen 3-nitrofluoranthene (3-NF). Although the specific molecular mechanisms involved in this difference in sensitivity for the induction of lung tumors in rats by 3,9-DNF
M C Consolo et al.
Mutation research, 210(2), 263-269 (1989-02-01)
The environmental pollutant 3-nitrofluoranthene is metabolized in vitro and in vivo to several products including the phenolic metabolites 3-nitrofluoranthen-6-ol (3NF-6-ol), 3-nitrofluoranthen-8-ol (3NF-8-ol), and 3-nitrofluoranthen-9-ol (3NF-9-ol). Similarly, 1-nitropyrene is metabolized to the phenolic metabolites 1-nitropyren-3-ol (1NP-3-ol), 1-nitropyren-6-ol (1NP-6-ol), and 1-nitropyren-8-ol (1NP-8-ol).
Genetic toxicology and carcinogenicity of mono- and dinitrofluoranthenes.
H Tokiwa et al.
Mutation research, 297(2), 181-195 (1993-09-01)
X Tang et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 35(3-4), 373-378 (1997-03-01)
When 21 vitamins including related compounds haemin, chlorophyllin, chlorophyll, biliverdin and bilirubin, as well as juices from five fruits and 25 vegetables and solvent extracts from the residues of fruits and vegetables were tested for their antimutagenic potencies with respect
J V Pothuluri et al.
Journal of toxicology and environmental health, 42(2), 209-218 (1994-06-01)
We investigated the metabolism of 3-nitrofluoranthene by filamentous fungus, Cunninghamella elegans ATCC 36112. Cunninghamella elegans metabolized about 72% of the 3-nitro[3,4-14C]fluoranthene added during 144 h of incubation to 2 major metabolites. These metabolites were separated by reversed-phase high-performance liquid chromatography

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service