Skip to Content
Merck
All Photos(1)

Key Documents

BCR309

6-Nitrochrysene

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H11NO2
CAS Number:
Molecular Weight:
273.29
Beilstein:
2134652
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

220 °C (dec.) (lit.)

format

neat

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1cc2c3ccccc3ccc2c4ccccc14

InChI

1S/C18H11NO2/c20-19(21)18-11-17-13-6-2-1-5-12(13)9-10-15(17)14-7-3-4-8-16(14)18/h1-11H

InChI key

UAWLTQJFZUYROA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

6-Nitrochrysene may be used as a certified reference material for the quantification of the analyte in air samples, standard reference material 1649a (urban dust), air and diesel particulate reference materials using gas chromatography-mass spectrometry with negative ion chemical ionization (GC/NICI-MS).

Analysis Note

For more information please see:
BCR309

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Muta. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

J Krzeminski et al.
Chemical research in toxicology, 13(11), 1143-1148 (2000-11-23)
The remarkable carcinogenic activity of 6-nitrochrysene (6-NC) in several animal models, and its environmental presence, suggest its potential importance with regard to human cancer development. Depending on the bioassay model, 6-NC can be activated by simple nitro reduction, ring oxidation
Karam El-Bayoumy et al.
Chemical research in toxicology, 15(7), 972-978 (2002-07-18)
Human exposure to the class of nitropolynuclear aromatic hydrocarbons is via inhalation and/or ingestion. Therefore, one of the goals of this study was to determine the propensity of the environmental contaminant 6-nitrochrysene (6-NC) for inducing mammary cancer following its oral
Y H Chae et al.
Cancer research, 56(9), 2052-2058 (1996-05-01)
The environmental contaminant 6-nitrochrysene (6-NC) has been shown to induce adenomas and adenocarcinomas in the colons of rats. The present study aimed at providing a better understanding of mechanisms that are responsible for this effect. Three female CD rats were
C R Valentine et al.
Environmental and molecular mutagenesis, 25(2), 85-96 (1995-01-01)
We have analyzed 41 mRNA-splicing mutants from the hypoxanthine-guanine phosphoribosyl-transferase (hprt) gene of Chinese hamster ovary (CHO) cells. Twenty-two of these mutants produced single cDNA PCR products with a partial or complete exon deletion; 19 mutants produced multiple cDNA PCR
R M Chen et al.
Archives of toxicology, 72(7), 395-401 (1998-08-26)
The present study has determined the effect of 6-nitrochrysene (6-NC) on hepatic and pulmonary cytochrome P450 (P450)-dependent monooxygenases using hamsters pretreated with the nitrated polycyclic aromatic hydrocarbon (nitro-PAH) at 5 mg/kg per day for 3 days. Pretreatment with 6-NC elevated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service