Skip to Content
Merck
All Photos(1)

Key Documents

BCR305

1-Nitropyrene

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H9NO2
CAS Number:
Molecular Weight:
247.25
Beilstein:
1882811
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

153-155 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc2ccc3cccc4ccc1c2c34

InChI

1S/C16H9NO2/c18-17(19)14-9-7-12-5-4-10-2-1-3-11-6-8-13(14)16(12)15(10)11/h1-9H

InChI key

ALRLPDGCPYIVHP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Nitronaphthalene, belonging to the class of nitrated-polycyclic aromatic hydrocarbons, is persistent in the environment. It is produced from direct sources such as diesel, gasoline exhaust and gas-phase reactions of polycyclic aromatic hydrocarbons (PAHs) with oxides of nitrogen.

Analysis Note

For more information please see:
BCR305

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ning Tang et al.
Chemosphere, 107, 324-330 (2014-02-11)
Airborne particulates were collected at a background site (Wajima Air Monitoring Station; WAMS) on the Noto Peninsula, Japan from January 2006 to December 2007. 1-, 2-nitropyrenes (1-, 2-NPs) and 2-nitrofluoranthene (2-NFR), in the particulates were determined with a sensitive HPLC
Takayuki Kameda et al.
Environmental science & technology, 45(8), 3325-3332 (2011-03-17)
The formation of hydroxynitropyrene (OHNP) via a photochemical reaction of 1-nitropyrene (1-NP) was demonstrated using a UV irradiation system. The photoreaction of 1-NP in methanol gave products that were hydroxy-substituted at position 1 and mononitro-substituted at positions 2, 3, 5
Wen-Cheng Chu et al.
Chemical research in toxicology, 22(12), 1938-1947 (2009-12-08)
Cytochrome P450 1a1 (Cyp1a1) is a phase I xenobiotic-metabolizing enzyme, the expression of which is mainly driven by the aryl hydrocarbon receptor (AhR). Cyp1a1 messenger (m)RNA is labile. Our study indicates that 1-nitropyrene (1-NP) highly induced Cyp1a1 protein expression, although
Justin P Miller-Schulze et al.
Journal of exposure science & environmental epidemiology, 23(2), 170-175 (2012-05-17)
Diesel exhaust (DE) is a significant contributor to the toxicity associated with particulate matter (PM). 1-Nitropyrene (1-NP) has been used as a molecular marker for DE, and the urinary metabolites of 1-NP have been proposed as biomarkers for exposure to
Jyan-Gwo J Su et al.
Toxicology, 244(2-3), 257-270 (2008-01-22)
It is reported that diesel exhaust particles contain more 1-nitropyrene (1-NP) than benzo[a]pyrene (B[a]P), both of which are potent carcinogenic compounds. In this study, we show that 1-NP is more potent in reducing cell viability than B[a]P, pyrene, nitrobenzene, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service