Skip to Content
Merck
All Photos(2)

Key Documents

89773

Supelco

Asiatic acid

analytical standard

Synonym(s):

2α,23-Dihydroxyursolic acid, Dammarolic acid, NSC 166063

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H48O5
CAS Number:
Molecular Weight:
488.70
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

325-330 °C (lit.)

application(s)

food and beverages

format

neat

SMILES string

C[C@@H]1CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)C[C@@H](O)[C@H](O)[C@@](C)(CO)[C@@H]5CC[C@@]34C)[C@@H]2[C@H]1C)C(O)=O

InChI

1S/C30H48O5/c1-17-9-12-30(25(34)35)14-13-28(5)19(23(30)18(17)2)7-8-22-26(3)15-20(32)24(33)27(4,16-31)21(26)10-11-29(22,28)6/h7,17-18,20-24,31-33H,8-16H2,1-6H3,(H,34,35)/t17-,18+,20-,21-,22-,23+,24+,26+,27+,28-,29-,30+/m1/s1

InChI key

JXSVIVRDWWRQRT-UYDOISQJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Asiatic acid (AA) has been used in a study to investigate its antihypertensive effect in high carbohydrate, high fat (HCHF) diet-induced MS metabolic syndrome (MS) rats. It has also been used in a study to understand its chemoprophylaxis effect against malaria in Sprague-Dawley rats.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Starting material for asiatic acid derivative synthesis for use as:
  • Anticancer agents
  • Glycogen phosphorylase inhibitors
  • Hepatoprotectants

Biochem/physiol Actions

Asiatic acid is commonly used in wound healing. Asiatic acid has antioxidant, anti-inflammatory and neuroprotective properties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pre-infection administration of asiatic acid retards parasitaemia induction in Plasmodium berghei murine malaria infected Sprague-Dawley rats.
Mavondo GA, et al.
Malaria Journal, 15(1), 1-1 (2016)
Asiatic acid attenuates renin-angiotensin system activation and improves vascular function in high-carbohydrate, high-fat diet fed rats
Putcharawipa M, et al.
BMC Complementary and Alternative Medicine, 16(1), 1-1 (2016)
Wenjie Guo et al.
International immunopharmacology, 24(2), 232-238 (2014-12-20)
In the present study, the effect of asiatic acid, a natural triterpenoid compound, on murine experimental colitis induced by dextran sulfate sodium (DSS) and its possible mechanism were examined in vivo and vitro. Oral administration of asiatic acid dose-dependently attenuated
Yongfang Yuan et al.
Journal of ethnopharmacology, 163, 31-38 (2015-01-18)
Asiatic acid is one of the main components in the herb Centella asiatica, which is a well-known herbal medicine for its excellent pharmacological effects. To enhance the development potentials of asiatic acid as a chemopreventative agent, there is a great

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service