Skip to Content
Merck
All Photos(1)

Key Documents

546712

Sigma-Aldrich

Asiatic acid

97%

Synonym(s):

2α,23-Dihydroxyursolic acid, Dammarolic acid, NSC 166063

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H48O5
CAS Number:
Molecular Weight:
488.70
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

325-330 °C (lit.)

functional group

carboxylic acid
hydroxyl

SMILES string

C[C@@H]1CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)C[C@@H](O)[C@H](O)[C@@](C)(CO)[C@@H]5CC[C@@]34C)[C@@H]2[C@H]1C)C(O)=O

InChI

1S/C30H48O5/c1-17-9-12-30(25(34)35)14-13-28(5)19(23(30)18(17)2)7-8-22-26(3)15-20(32)24(33)27(4,16-31)21(26)10-11-29(22,28)6/h7,17-18,20-24,31-33H,8-16H2,1-6H3,(H,34,35)/t17-,18+,20-,21-,22-,23+,24+,26+,27+,28-,29-,30+/m1/s1

InChI key

JXSVIVRDWWRQRT-UYDOISQJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Starting material for asiatic acid derivative synthesis for use as:
  • Anticancer agents
  • Glycogen phosphorylase inhibitors
  • Hepatoprotectants

Biochem/physiol Actions

Asiatic acid is commonly used in wound healing. Asiatic acid has antioxidant, anti-inflammatory and neuroprotective properties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kaudjhis Patrick Ulrich N'deh et al.
Nanomaterials (Basel, Switzerland), 10(3) (2020-03-19)
Biocompatibility is very important for cell growth using 3D printers, but biocompatibility materials are very expensive. In this study, we investigated the possibility of cell culture by the surface modification of relatively low-cost industrial materials and an efficient three-dimensional (3D)
Chandagirikoppal V Kavitha et al.
PloS one, 6(8), e22745-e22745 (2011-08-10)
Malignant gliomas are one of the most devastating and incurable tumors. Sustained excessive angiogenesis by glioma cells is the major reason for their uncontrolled growth and resistance toward conventional therapies resulting in high mortality. Therefore, targeting angiogenesis should be a
Wenjie Guo et al.
PloS one, 7(9), e46018-e46018 (2012-10-03)
Selectively facilitating apoptosis of activated T cells is essential for the clearance of pathogenic injurious cells and subsequent efficient resolution of inflammation. However, few chemicals have been reported to trigger apoptosis of activated T cells for the treatment of hepatitis
M N Nasir et al.
Journal of ethnopharmacology, 134(2), 203-209 (2010-12-21)
Centella asiatica has a reputation to restore declining cognitive function in traditional medicine. To date, only a few compounds that show enhancing learning and memory properties are available. Therefore, the present study investigates the effects of for acute administration of
M N Nasir et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 19(3-4), 311-316 (2011-11-25)
The asiatic acid, a triterpenoids isolated from Centella asiatica was used to delineate its inhibitory effect on acetylcholinesterase (AChE) properties, excitatory post synaptic potential (EPSP) and locomotor activity. This study is consistent with asiatic acid having an effect on AChE

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service