Skip to Content
Merck
All Photos(1)

Key Documents

43191

Supelco

Asiaticoside

analytical standard

Synonym(s):

Madecassol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H78O19
CAS Number:
Molecular Weight:
959.12
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

[H][C@]12CC=C3C4[C@@H](C)[C@H](C)CC[C@@]4(CC[C@@]3(C)[C@]1(C)CCC5[C@](C)(CO)[C@@H](O)[C@H](O)C[C@]25C)C(=O)O[C@@H]6O[C@H](CO[C@@H]7O[C@H](CO)[C@@H](O[C@@H]8O[C@H](C)[C@H](O)[C@@H](O)[C@H]8O)[C@H](O)[C@H]7O)[C@@H](O)[C@H](O)[C@H]6O

InChI

1S/C48H78O19/c1-20-10-13-48(15-14-46(6)23(29(48)21(20)2)8-9-28-44(4)16-24(51)39(60)45(5,19-50)27(44)11-12-47(28,46)7)43(61)67-42-36(58)33(55)31(53)26(65-42)18-62-40-37(59)34(56)38(25(17-49)64-40)66-41-35(57)32(54)30(52)22(3)63-41/h8,20-22,24-42,49-60H,9-19H2,1-7H3/t20-,21+,22-,24-,25-,26-,27?,28-,29?,30+,31-,32-,33+,34-,35-,36-,37-,38-,39+,40-,41+,42+,44+,45+,46-,47-,48+/m1/s1

InChI key

WYQVAPGDARQUBT-TZXSXNIFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fatma J Al-Saeedi et al.
Journal of nuclear medicine technology, 39(4), 279-283 (2011-11-15)
This study was done to examine the effect of asiaticoside on MCF-7 cell uptake of (99m)Tc-tetrofosmin ((99m)Tc-Tfos) and (99m)Tc-sestamibi ((99m)Tc-MIBI). The 3-(4,5-dimethylthiozol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) assay was used to evaluate the effect of a 50% inhibitory concentration of asiaticoside on MCF-7
Ipek Süntar et al.
Journal of ethnopharmacology, 132(2), 408-413 (2010-08-17)
The leaves, flowers, and whole aerial parts of Verbascum L. (Scrophulariaceae) species are used to treat eczema and other types of inflammatory skin conditions and as a desiccant for wounds in Turkish traditional medicine. In the present study, the methanolic
Siddhartha Kumar Bhaumik et al.
The Journal of antimicrobial chemotherapy, 67(4), 910-920 (2012-01-20)
The aim of this study was to investigate and characterize the efficacy of asiaticoside in an experimental model of visceral leishmaniasis caused by antimony-susceptible (AG83) and -resistant (GE1F8R and K39) Leishmania donovani. The effect of asiaticoside was evaluated by microscopic
Jisha Satheesan et al.
Mycorrhiza, 22(3), 195-202 (2011-06-21)
Centella asiatica (Indian pennywort) has wide application in Indian and Chinese traditional medicines with documented evidence for wound healing and neuroprotective and anti-aging potential. Asiaticoside, a trisaccharide triterpene, is the most medicinally active compound in the plant. β-Amyrin synthase and
Jiajia Zheng et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 35(11), 1482-1485 (2010-09-09)
The present study is to investigate the protective effects of asiaticoside on sepsis-induced acute kidney injury in mice. With the sepsis induced by cecal ligation and puncture (CLP), forty eight kunming mice were randomly divided into four groups as sham

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service