Skip to Content
Merck
All Photos(1)

Documents

79266

Supelco

Trimethylphenylammonium hydroxide solution

~0.5 M (CH3)3N(OH)C6H5 in methanol, for GC derivatization, LiChropur

Synonym(s):

Phenyltrimethylammonium hydroxide, TMAH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N(OH)C6H5
CAS Number:
Molecular Weight:
153.22
Beilstein:
3917033
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for GC derivatization

Quality Level

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Esterifications

concentration

~0.5 M (CH3)3N(OH)C6H5 in methanol

technique(s)

gas chromatography (GC): suitable

impurities

≤0.2% halides (as chloride)

storage temp.

2-8°C

SMILES string

[OH-].C[N+](C)(C)c1ccccc1

InChI

1S/C9H14N.H2O/c1-10(2,3)9-7-5-4-6-8-9;/h4-8H,1-3H3;1H2/q+1;/p-1

InChI key

HADKRTWCOYPCPH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Trimethylphenylammonium hydroxide (TMAH) is a methylating reagent.

Application

Learn more in the Product Information
Suitable for the derivatization of amino acids, n-methyl and n-aryl carbamates and fatty acids, clonidine, and substituted phenylureas.
TMAH may be used as a 0.1 mole/litre solution in methanol to determine plasma concentrations of carbamazepine and other anticonvulsant drugs, including phenobarbital, diphenylhydantoin, primidone, and mephenytoin using Gas-Liquid Chromatography.

Other Notes

Reagent for n-methyl and methyl esters.
Sales restrictions may apply

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of carbamazapine ("Tegretol") and other anticonvulsants in human plasma by gas-liquid chromatography.
J C Roger et al.
Clinical chemistry, 19(6), 590-592 (1973-06-01)
A Ranz et al.
Journal of biochemical and biophysical methods, 69(1-2), 3-14 (2006-03-28)
In the present study, a derivatization method for the determination of acidic herbicides has been investigated. This procedure involves the methylation with the quaternary ammonium salt trimethylanilinium hydroxide (TMAH) directly in the gas chromatographic auto-sampler vial for analysis by gas
Timothy J Jensen et al.
Journal of photochemistry and photobiology. B, Biology, 100(2), 100-111 (2010-06-19)
Five cationic porphyrins bearing one to four -N(CH(3))(3)(+) groups linked to the p-phenyl positions of 5,10,15,20-tetraphenylporphyrin (TPP) were synthesized in order to study the effect of overall charge and its distribution on the cellular uptake, phototoxicity and intracellular localization using
Jie Zhang et al.
Journal of chromatography. A, 1216(44), 7527-7532 (2009-04-07)
A method has been established for the determination of four pharmaceutically active compounds (ibuprofen, ketoprofen, naproxen and clofibric acid) in water samples using dynamic hollow fiber liquid-phase microextraction (HF/LPME) followed by gas chromatography (GC) injection port derivatization and GC-mass spectrometric
Alec N Salt et al.
Hearing research, 283(1-2), 14-23 (2011-12-20)
It has been widely believed that drug entry from the middle ear into perilymph occurs primarily via the round window (RW) membrane. Entry into scala vestibuli (SV) was thought to be dominated by local, inter-scala communication between scala tympani (ST)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service