Merck
  • Marker entry into vestibular perilymph via the stapes following applications to the round window niche of guinea pigs.
Marker entry into vestibular perilymph via the stapes following applications to the round window niche of guinea pigs.
Hearing research (2011-12-20)
Alec N Salt, Elisha B King, Jared J Hartsock, Ruth M Gill, Stephen J O'Leary
ABSTRACT
It has been widely believed that drug entry from the middle ear into perilymph occurs primarily via the round window (RW) membrane. Entry into scala vestibuli (SV) was thought to be dominated by local, inter-scala communication between scala tympani (ST) and SV through permeable tissues such as the spiral ligament. In the present study, the distribution of the ionic marker trimethylphenylammonium (TMPA) was compared following intracochlear injections or applications to the RW niche, with or without occlusion of the RW membrane or stapes area. Perilymph TMPA concentrations were monitored either in real time with TMPA-selective microelectrodes sealed into ST and SV, or by the collection of sequential perilymph samples from the lateral semi-circular canal. Local inter-scala communication of TMPA was confirmed by measuring SV and ST concentrations following direct injections into perilymph of ST. Application of TMPA to the RW niche also showed a predominant entry into ST, with distribution to SV presumed to occur secondarily. When the RW membrane was occluded by a silicone plug, RW niche irrigation produced higher concentrations in SV compared to ST, confirming direct TMPA entry into the vestibule in the region of the stapes. The proportion of TMPA entering by the two routes was quantified by perilymph sampling from the lateral semi-circular canal. The TMPA levels of initial samples (originating from the vestibule) were markedly lower when the stapes area was occluded with silicone. These data were interpreted using a simulation program that incorporates all the major fluid and tissue compartments of the cochlea and vestibular systems. From this analysis it was estimated that 65% of total TMPA entered through the RW membrane and 35% entered the vestibule directly in the vicinity of the stapes. Direct entry of drugs into the vestibule is relevant to inner ear fluid pharmacokinetics and to the growing field of intratympanic drug delivery.
Sigma-Aldrich
Trimethylphenylammonium bromide, 98%
Sigma-Aldrich
Trimethylphenylammonium chloride, ≥98%
Supelco
Trimethylphenylammonium hydroxide solution, ~0.5 M (CH3)3N(OH)C6H5 in methanol, for GC derivatization, LiChropur
Sigma-Aldrich
Trimethylphenylammonium hydroxide solution, ~25% in H2O (1.68 M)
Sigma-Aldrich
Trimethylphenylammonium tribromide, 97%