Skip to Content
Merck
All Photos(4)

Key Documents

H4515

Sigma-Aldrich

Methyl heptadecanoate

≥99%

Synonym(s):

Heptadecanoic acid methyl ester, Methyl margarate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15COOCH3
CAS Number:
Molecular Weight:
284.48
Beilstein:
1782681
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Assay

≥99%

form

powder or solid

bp

152-153 °C/0.05 mmHg (lit.)

mp

29.8-30.3 °C (lit.)

functional group

ester

lipid type

saturated FAs

shipped in

ambient

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCC(=O)OC

InChI

1S/C18H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20-2/h3-17H2,1-2H3

InChI key

HUEBIMLTDXKIPR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl heptadecanoate has been used:
  • as a substrate for lipoxygenase assay
  • as an internal standard for the quantification of squalene, fatty acids (FA) and lipids
  • as a reference standard for precise separation of methyl esters of different fatty acids

Other Notes

Product clings due to static, may require dissolving material to fully recover out of bottle.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipolytic and oxidative changes of barley lipids during malting and mashing
Kaukovirta-Norja A, et al.
Journal of the Institute of Brewing, 99(5), 395-403 (1993)
The isolation and characterization of Pseudozyma sp. JCC 207, a novel producer of squalene
Chang MH, et al.
Applied Microbiology and Biotechnology, 78(6), 963-963 (2008)
Effect of dairy calcium on fecal fat excretion: a randomized crossover trial
Bendsen NT, et al.
International Journal of Obesity, 32(12), 1816-1816 (2008)
Importance of the fat content within the cheese-matrix for blood lipid profile, faecal fat excretion, and gut microbiome in growing pigs
Thorning TK, et al.
International dairy journal, 61, 67-75 (2016)
Effects of docosahexaenoic Acid supplementation on blood pressure, heart rate, and serum lipids in Scottish men with hypertension and hypercholesterolemia
Sagara M, et al.
International Journal of Hypertension, 2011 (2011)

Protocols

Protocol for GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on Equity®-1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service