Skip to Content
Merck
All Photos(1)

Key Documents

72336

Supelco

N,N-Dimethylacetamide

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CON(CH3)2
CAS Number:
Molecular Weight:
87.12
Beilstein:
1737614
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3 (vs air)

vapor pressure

2 mmHg ( 25 °C)
4 mmHg ( 38 °C)

Assay

≥99.9% (GC)

autoignition temp.

914 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

1.8 %, 100 °F
11.5 %, 160 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.437 (lit.)

bp

164.5-166 °C (lit.)

mp

−20 °C (lit.)

density

0.937 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CN(C)C(C)=O

InChI

1S/C4H9NO/c1-4(6)5(2)3/h1-3H3

InChI key

FXHOOIRPVKKKFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethylacetamide is a highly polar solvent, which can find applications in a variety of chemical reactions. A mixture of lithium chloride in N,N-dimethylacetamide may be used for the dissolution of cellulose to form a homogeneous solution.

Application

N,N-Dimethylacetamide may be used as an analytical standard for the determination of the analyte in pharmaceuticals, biological samples, water and air samples by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solvent effects on the intramolecular charge transfer character of N, N-diaryl dihydrophenazine catalysts for organocatalyzed atom transfer radical polymerization
Ryan DM, eta l.
Journal of Polymer Science Part A: Polymer Chemistry (2017)
Lucia L and Ayoub A
Polysaccharide-based Fibers and Composites: Chemical and Engineering Fundamentals and Industrial Applications (2017)
Separate determination by gas-chromatography of dimethylformamide, dimethylacetamide, monomethylformamide and monomethylacetamide in urine for biological monitoring
Kawai T, et al.
Journal of Occupational Health, 39(2), 113-118 (1997)
Gas chromatography/mass spectrometric determination of traces of hydrophilic and volatile organic compounds in water after preconcentration with activated carbon
KADOKAMI K, et al.
Analytical Sciences, 6(6), 843-849 (1990)
Simultaneous Determination of Dimethylformamide and N, N-Dimethylacetamide in Air by Capillary Gas Chromatography [J]
Xiao-Juan, LI
Chinese Journal of Spectroscopy Laboratory, 3 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service