Skip to Content
Merck
All Photos(1)

Key Documents

522414

Sigma-Aldrich

N,N-Dimethylacetamide-d9

99 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CON(CD3)2
CAS Number:
Molecular Weight:
96.18
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

99 atom % D

Quality Level

Assay

99% (CP)

form

liquid

technique(s)

NMR: suitable

refractive index

n20/D 1.438 (lit.)

bp

164-166 °C (lit.)

mp

−20 °C (lit.)

density

1.033 g/mL at 25 °C

mass shift

M+9

SMILES string

O=C(N(C([2H])([2H])[2H])C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C4H9NO/c1-4(6)5(2)3/h1-3H3/i1D3,2D3,3D3

InChI key

FXHOOIRPVKKKFG-GQALSZNTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethylacetamide-d9 (DMA-d9) is a deuterated analog of N, N-dimethylacetamide. It has an isotopic purity of 99 atom % D. It is a standard purity solvent suitable for routine NMR analyses.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inês Farinha et al.
Carbohydrate polymers, 130, 455-464 (2015-06-17)
Purified chitin-glucan complex (CGCpure) was extracted from Komagataella pastoris biomass using a hot alkaline treatment, followed by neutralization and repeated washing with deionized water. The co-polymer thus obtained had a β-glucan:chitin molar ratio of 75:25 and low protein and inorganic
Tony Reid et al.
The Lancet. Oncology, 16(9), 1133-1142 (2015-08-25)
Epigenetic alterations have been strongly associated with tumour formation and resistance to chemotherapeutic drugs, and epigenetic modifications are an attractive target in cancer research. RRx-001 is activated by hypoxia and induces the generation of reactive oxygen and nitrogen species that
Fatma Kayaci et al.
Nanoscale, 6(17), 10224-10234 (2014-07-25)
Oxygen vacancies (V(O)s) in ZnO are well-known to enhance photocatalytic activity (PCA) despite various other intrinsic crystal defects. In this study, we aim to elucidate the effect of zinc interstitials (Zn(i)) and V(O)s on PCA, which has applied as well
Takayuki Negishi et al.
Neurotoxicology and teratology, 44, 46-52 (2014-06-03)
Bisphenol A (BPA) is a widespread environmental contaminant, and humans are routinely exposed to BPA. We investigated whether prenatal exposure to BPA influences behavioral development in juvenile cynomolgus monkeys (Macaca fascicularis). Pregnant cynomolgus monkeys were implanted with subcutaneous pumps and
Naoki Aizawa et al.
Neurourology and urodynamics, 34(4), 368-374 (2014-02-18)
We measured single-unit mechanosensitive afferent activities (SAAs) during reflexic, rhythmic bladder contractions (RBCs), and examined whether L-arginine, an NO substrate, and mirabegron, a β3-adrenoceptor agonist, and oxybutynin, an antimuscarinic agent, can affect the SAAs in such condition. Twenty-nine female Sprague-Dawley

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service