Skip to Content
Merck
All Photos(1)

Key Documents

5.89582

Sigma-Aldrich

N,N-Dimethylacetamide

99.8%, anhydrous

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CON(CH3)2
CAS Number:
Molecular Weight:
87.12
Beilstein:
1737614
MDL number:
UNSPSC Code:
12352111
NACRES:
NA.22

grade:
anhydrous
bp:
164.5-166 °C (lit.)
vapor pressure:
2 mmHg ( 25 °C)
4 mmHg ( 38 °C)
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

vapor density

3 (vs air)

vapor pressure

2 mmHg ( 25 °C)
4 mmHg ( 38 °C)

Assay

99.8%

autoignition temp.

914 °F

expl. lim.

1.8 %, 100 °F
11.5 %, 160 °F

refractive index

n20/D 1.437 (lit.)

bp

164.5-166 °C (lit.)

mp

−20 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service