Skip to Content
Merck
All Photos(1)

Key Documents

68320

Supelco

3-Methylpentane

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)2CHCH3
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730734
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor density

2.97 (vs air)

vapor pressure

135 mmHg ( 17 °C)

Assay

≥99.5% (GC)

autoignition temp.

532 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

~7.7 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.376 (lit.)
n20/D 1.376

bp

64 °C (lit.)

density

0.664 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCC(C)CC

InChI

1S/C6H14/c1-4-6(3)5-2/h6H,4-5H2,1-3H3

InChI key

PFEOZHBOMNWTJB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

19.4 °F - closed cup

Flash Point(C)

-7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Richard C Bell et al.
The Journal of chemical physics, 127(2), 024704-024704 (2007-07-21)
An ion soft-landing approach was applied to study the motion of hydronium (D(3)O(+)) and cesium (Cs(+)) ions from 84 to 104 K in glassy 3-methyl-pentane (3MP) films vapor deposited on Pt(111). Both ions were found to have very similar mobilities
3-Methylpentane. CAS# 96-14-0.
J B Galvin et al.
Journal of toxicology and environmental health. Part A, 58(1-2), 93-102 (1999-10-28)
R Hutterer et al.
Biochimica et biophysica acta, 1323(2), 195-207 (1997-01-31)
Time-resolved fluorescence measurements were performed for a set of n-anthroyloxy fatty acids (n-AS; n = 2, 3, 6, 9, 12, 16) in both solvent and vesicle systems. The Stokes' shifts and the mean relaxation times calculated from the time-resolved emission
M W Grinstaff et al.
Science (New York, N.Y.), 264(5163), 1311-1313 (1994-05-27)
Halogenation of an iron porphyrin causes severe saddling of the macrocyclic structure and a large positive shift in the iron(III)/(II) redox couple. Although pre-halogenated iron(II) porphyrins such as Fe(TFPPBr8) [H2TFPPBr8, beta-octabromo-tetrakis(pentafluorophenyl)-porphyrin] are relatively resistant to autoxidation, they rapidly reduce alkyl
Shervin Shahriari et al.
The Journal of chemical physics, 121(18), 8960-8967 (2004-11-06)
We have measured the dielectric relaxation of several glass forming branched alkanes with very low dielectric loss in the frequency range 50 Hz-20 kHz. The molecular liquids of this study are 3-methylpentane, 3-methylheptane, 4-methylheptane, 2,3-dimethylpentane, and 2,4,6-trimethylheptane. All liquids display

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service