68312
2-Methylpentane
≥95.0% (GC)
Synonym(s):
‘Isohexane’
About This Item
vapor density
3 (vs air)
Quality Level
vapor pressure
6.77 psi ( 37.7 °C)
Assay
≥95.0% (GC)
form
liquid
autoignition temp.
583 °F
expl. lim.
7 %
impurities
≤0.5% 3-methylpentane
≤2% 2,3-dimethyl-propane
refractive index
n20/D 1.371 (lit.)
n20/D 1.371
bp
62 °C (lit.)
mp
−154 °C (lit.)
density
0.653 g/mL at 25 °C (lit.)
SMILES string
CCCC(C)C
InChI
1S/C6H14/c1-4-5-6(2)3/h6H,4-5H2,1-3H3
InChI key
AFABGHUZZDYHJO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
19.4 °F - closed cup
Flash Point(C)
-7 °C - closed cup
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service