Skip to Content
Merck
All Photos(2)

Key Documents

50202

Supelco

Tetrahydrocurcumin

analytical standard

Synonym(s):

1,7-Bis(4-hydroxy-3-methoxyphenyl)-3,5-heptanedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24O6
CAS Number:
Molecular Weight:
372.41
Beilstein:
3485469
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

COc1cc(CCC(=O)CC(=O)CCc2ccc(O)c(OC)c2)ccc1O

InChI

1S/C21H24O6/c1-26-20-11-14(5-9-18(20)24)3-7-16(22)13-17(23)8-4-15-6-10-19(25)21(12-15)27-2/h5-6,9-12,24-25H,3-4,7-8,13H2,1-2H3

InChI key

LBTVHXHERHESKG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrahydrocurcumin is a colorless compound and a major metabolite of curcumin, which is known to possess antioxidant activity and inhibitory effects on tertiary butyl hydroperoxide-induced lipid peroxidation. It can be obtained via hydrogenation of curcumin in the presence of platinum(IV)oxide.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: curcuma

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

385.2 °F

Flash Point(C)

196.2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kei Shimoda et al.
Natural product communications, 7(4), 529-530 (2012-05-12)
Cultured plant cells of Marchantia polymorpha, Nicotiana tabacum, Phytolacca americana, Catharanthus roseus, and Gossypium hirsutum were examined for their ability to reduce curcumin. Only M. polymorpha cells converted curcumin into tetrahydrocurcumin in 90% yield in one day. Time-course experiment revealed
Symone M San Miguel et al.
Archives of oral biology, 56(8), 812-822 (2011-04-05)
Antioxidants (AOs) are the first line of defence against free radical damage and are critical for maintaining optimum health and well being. The need for AOs becomes even more critical with increased exposure to free radicals generated by pollution, cigarette
Ching-Shu Lai et al.
Molecular nutrition & food research, 55(12), 1819-1828 (2011-09-03)
Tetrahydrocurcumin (THC), a major metabolite of curcumin (CUR), has been demonstrated to be anti-cancerogenic and anti-angiogenic and prevents type II diabetes. In this present study, we investigated the chemopreventive effects and underlying molecular mechanisms of dietary administration of CUR and
Pravit Asawanonda et al.
Photomedicine and laser surgery, 28(5), 679-684 (2010-10-22)
The aim of this study was to compare the efficacy of targeted narrowband UVB phototherapy plus topical tetrahydrocurcuminoid with that of targeted narrowband UVB monotherapy for induction of repigmentation in vitiligo. The 308-nm excimer laser and targeted narrowband UVB phototherapy
Symone M San Miguel et al.
Journal of periodontology, 81(11), 1675-1690 (2010-07-20)
Smoking is associated with an increased risk of oral health and dental problems. The aim of this study is to address the hypothesis that nicotine impairs wound healing by increasing reactive oxygen species and inhibiting cell migration, and antioxidants (AOs)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service