Skip to Content
Merck
All Photos(1)

Key Documents

46437

Supelco

Isoxaflutole

PESTANAL®, analytical standard

Synonym(s):

5-Cyclopropyl-4-[2-(methylsulfonyl)-4-(trifluoromethyl)benzoyl]isoxazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12F3NO4S
CAS Number:
Molecular Weight:
359.32
Beilstein:
8344543
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CS(=O)(=O)c1cc(ccc1C(=O)c2cnoc2C3CC3)C(F)(F)F

InChI

1S/C15H12F3NO4S/c1-24(21,22)12-6-9(15(16,17)18)4-5-10(12)13(20)11-7-19-23-14(11)8-2-3-8/h4-8H,2-3H2,1H3

InChI key

OYIKARCXOQLFHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isoxaflutole is a novel pre-emergence herbicide widely employed for the protection of corn and sugarcane against broadleaf and grass weeds. Its mode of action involves the facilitation of characteristic foliar bleaching activity in target species preceding the plant death.

Application

Isoxaflutole may be used as an analytical reference standard for the determination of the analyte in blackcurrants and soya grain by gas chromatography-tandem mass spectrometry (GC-MS/MS) and liquid chromatography-tandem mass spectrometry (LC-MS/MS), respectively.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Sharon K Papiernik et al.
Journal of agricultural and food chemistry, 55(21), 8630-8639 (2007-09-21)
Two-year field dissipation studies were conducted in three soil types in Minnesota to examine the processes affecting the dissipation of the herbicide isoxaflutole and its phytotoxic diketonitrile metabolite (DKN) under relatively cool, wet soil conditions. Plots of cuphea were treated
K E Pallett et al.
Pest management science, 57(2), 133-142 (2001-07-18)
This paper reviews the discovery of isoxaflutole (IFT), focusing on the chemical and physicochemical properties which contribute to the herbicidal behaviour of this new herbicide. IFT (5-cyclopropyl-1,2-isoxazol-4-yl alpha alpha alpha-trifluoro-2-mesyl-p-tolyl ketone) is a novel herbicide for pre-emergence control of a
R N Lerch et al.
Journal of agricultural and food chemistry, 55(5), 1893-1899 (2007-02-08)
Isoxaflutole (IXF; Balance) belongs to a new class of isoxazole herbicides. Isoxaflutole has a very short half-life in soil and rapidly degrades to a stable and phytotoxic degradate, diketonitrile (DKN). DKN was previously discovered to rapidly react with hypochlorite (OCl-)
Pamela J Rice et al.
Journal of agricultural and food chemistry, 52(25), 7621-7627 (2005-01-29)
The transformation of isoxaflutole (ISOX) to its herbicidally active diketonitrile degradate (DKN) was significantly enhanced in the presence of soil and occurred more rapidly in systems containing soil with a greater soil pH. Sorption-desorption of ISOX and DKN in five
Validation and use of a QuEChERS-based gas chromatographic-tandem mass spectrometric method for multiresidue pesticide analysis in blackcurrants including studies of matrix effects and estimation of measurement uncertainty.
Walorczyk S.
Talanta, 120(2), 106-113 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service