Skip to Content
Merck
All Photos(1)

Key Documents

36978

Supelco

Pesticide standard 17 solution

PESTANAL®, analytical standard, according to DIN 38407, in acetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Agency

according to DIN 38407

product line

PESTANAL®

quality

in acetonitrile

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

multi-component solution

storage temp.

2-8°C

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Components

10 μg/mL each: atrazin, atrazin-desethyl, cyanazin, sebuthylazin, simazin, terbuthylazin, hexazinon, chlortoluron, diuron, isoproturon, linuron, methabenzthiazuron, metobromuron, metoxuron, monolinuron, metazoachlor, metolachlor

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Britany Rufenach et al.
Structure (London, England : 1993), 28(8), 922-932 (2020-06-04)
STAC3 is a soluble protein essential for skeletal muscle excitation-contraction (EC) coupling. Through its tandem SH3 domains, it interacts with the cytosolic II-III loop of the skeletal muscle voltage-gated calcium channel. STAC3 is the target for a mutation (W284S) that
Nahid Iglesias et al.
Molecular cell, 77(1), 51-66 (2019-12-01)
Spatially and functionally distinct domains of heterochromatin and euchromatin play important roles in the maintenance of chromosome stability and regulation of gene expression, but a comprehensive knowledge of their composition is lacking. Here, we develop a strategy for the isolation of
William T Yewdell et al.
Immunity, 53(5), 952-970 (2020-10-26)
Precise targeting of activation-induced cytidine deaminase (AID) to immunoglobulin (Ig) loci promotes antibody class switch recombination (CSR) and somatic hypermutation (SHM), whereas AID targeting of non-Ig loci can generate oncogenic DNA lesions. Here, we examined the contribution of G-quadruplex (G4)
Boyuan Wang et al.
Molecular cell, 80(1), 29-42 (2020-08-29)
(p)ppGpp is a nucleotide messenger universally produced in bacteria following nutrient starvation. In E. coli, ppGpp inhibits purine nucleotide synthesis by targeting several different enzymes, but the physiological significance of their inhibition is unknown. Here, we report the structural basis of

Articles

Pesticides analysis by LC-MS needs pesticide-free reagent water, which can be reliably delivered by Milli-Q® IQ 7 ultrapure water systems. See data.

Pesticides analysis by LC-MS needs pesticide-free reagent water, which can be reliably delivered by Milli-Q® IQ 7 ultrapure water systems. See data.

Pesticides analysis by LC-MS needs pesticide-free reagent water, which can be reliably delivered by Milli-Q® IQ 7 ultrapure water systems. See data.

Pesticides analysis by LC-MS needs pesticide-free reagent water, which can be reliably delivered by Milli-Q® IQ 7 ultrapure water systems. See data.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service