Skip to Content
Merck
All Photos(1)

Key Documents

36173

Supelco

Monocrotophos

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14NO5P
CAS Number:
Molecular Weight:
223.16
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CNC(=O)\C=C(/C)OP(=O)(OC)OC

InChI

1S/C7H14NO5P/c1-6(5-7(9)8-2)13-14(10,11-3)12-4/h5H,1-4H3,(H,8,9)/b6-5+

InChI key

KRTSDMXIXPKRQR-AATRIKPKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Monocrotophos is a broad spectrum organophosphate pesticide, which can be commonly used for agricultural purposes.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Muta. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 4

1 of 4

Biochemical alteration induced by monocrotophos in the blood plasma of fish, Channa punctatus (Bloch)
Agrahari S, et al.
Pesticide Biochemistry and Physiology, 88(3), 268-272 (2007)
Sonophotocatalytic degradation of monocrotophos using TiO 2 and Fe 3+
Madhavan J, et al.
Journal of Hazardous Materials, 177(1), 944-949 (2010)
A novel acetylcholinesterase biosensor based on chitosan-gold nanoparticles film for determination of monocrotophos using FFT continuous cyclic voltammetry
Norouzi.P,et al.
International Journal of Electrochemical Science, 5, 1434- 1446 (2010)
Deok Ho Kwon et al.
Insect biochemistry and molecular biology, 42(3), 212-219 (2011-12-27)
The mutations (G228S, A391T and F439W) and duplication of the acetylcholinesterase (AChE) gene (Tuace) are involved in monocrotophos resistance in the two-spotted spider mites, Tetranychus urticae (Kwon et al., 2010a, b). The overexpression of T. urticae AChE (TuAChE) as a result of
Hua Tian et al.
Toxicology and applied pharmacology, 263(2), 163-170 (2012-06-21)
Monocrotophos is a highly toxic organophosphorus pesticide that has been confirmed to be an endocrine-disrupting chemical. To evaluate the influence of this pollutant on the reproductive system of male fish, we studied the sex steroid levels, reproductive traits, sex ratio

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service