Skip to Content
Merck
All Photos(1)

Key Documents

46277

Supelco

Esfenvalerate

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H22ClNO3
CAS Number:
Molecular Weight:
419.90
Beilstein:
4275674
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C)[C@H](C(=O)O[C@H](C#N)c1cccc(Oc2ccccc2)c1)c3ccc(Cl)cc3

InChI

1S/C25H22ClNO3/c1-17(2)24(18-11-13-20(26)14-12-18)25(28)30-23(16-27)19-7-6-10-22(15-19)29-21-8-4-3-5-9-21/h3-15,17,23-24H,1-2H3/t23-,24+/m1/s1

InChI key

NYPJDWWKZLNGGM-RPWUZVMVSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Esfenvalerate may be used as an analytical reference standard for the quantification of the analyte in environmental samples using gas chromatography-negative ion chemical ionization mass spectrometry (GC-NCI-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2 - STOT SE 1

Target Organs

Nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthias Liess et al.
Scientific reports, 9(1), 15248-15248 (2019-10-28)
We present a model to identify the effects of low toxicant concentrations. Due to inadequate models, such effects have so far often been misinterpreted as random variability. Instead, a tri-phasic relationship describes the effects of a toxicant when a broad
John L Maron et al.
Ecology, 99(2), 464-473 (2017-12-06)
Many clonal organisms occasionally outcross, but the long-term consequences of such infrequent events are often unknown. During five years, representing three to five plant generations, we followed 16 experimental field populations of the forb, Oenothera biennis, originally planted with the same
Mark G Teese et al.
PloS one, 8(6), e65951-e65951 (2013-06-27)
Esterases have recurrently been implicated in insecticide resistance in Helicoverpa armigera but little is known about the underlying molecular mechanisms. We used a baculovirus system to express 14 of 30 full-length esterase genes so far identified from midgut cDNA libraries
Nedim Tüzün et al.
Environmental science & technology, 51(23), 13949-13956 (2017-11-08)
Current ecological risk assessment of pesticides fails to protect aquatic biodiversity. For the first time, we tested two potential reasons for this failure with regard to carry-over effects across metamorphosis: their dependence on hatching period, and the lack of studies
Khuong Van Dinh et al.
Evolutionary applications, 9(3), 450-461 (2016-03-19)
Many species are too slow to track their poleward-moving climate niche under global warming. Pesticide exposure may contribute to this by reducing population growth and impairing flight ability. Moreover, edge populations at the moving range front may be more vulnerable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service