Skip to Content
Merck
All Photos(2)

Key Documents

36515

Supelco

Mecarbam

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20NO5PS2
CAS Number:
Molecular Weight:
329.37
Beilstein:
1888011
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOC(=O)N(C)C(=O)CSP(=S)(OCC)OCC

InChI

1S/C10H20NO5PS2/c1-5-14-10(13)11(4)9(12)8-19-17(18,15-6-2)16-7-3/h5-8H2,1-4H3

InChI key

KLGMSAOQDHLCOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neuropathy following acute intoxication with Mecarbam (OP ester)
N Senanayake
Acta neurologica Scandinavica, 85(1), 77-77 (1992-01-01)
E Stamboulis et al.
Acta neurologica Scandinavica, 83(3), 198-200 (1991-03-01)
Only a small number of organophosphorous compounds, of the many thousands circulating on the market, has been reported as causing neuropathy with delayed onset. A case is presented of a young male who in an attempt to commit suicide by
R S Greenberg
Journal - Association of Official Analytical Chemists, 70(5), 870-874 (1987-09-01)
A gas chromatographic method is described for determining residues of mecarbam and 3 of its metabolites, mecarboxon, diethoate, and diethoxon, in cottonseeds. For mecarbam analysis, following Soxhlet extraction with chloroform (after blending), the oily extract is partitioned with propylene carbonate
H Tsoukali-Papadopoulou et al.
Forensic science international, 35(4), 249-252 (1987-12-01)
A suicidal poisoning committed by a 19-year-old pregnant woman, who ingested an unknown quantity of an aqueous solution mecarbam (trade name includes 'Murfotox') containing 68 g/l of S-(N-ethoxycarbonyl-N-methylcarbamoyl methyl) O,O-diethyl phosphorodithioate (C.R. Worthing (Ed.), The Pesticide Manual, 6th end, BCPC
A M Tsatsakis et al.
Veterinary and human toxicology, 38(2), 101-107 (1996-04-01)
Nine human acute poisonings due to intentional ingestion of organophosphorous pesticides are presented. Six of the victims died. Six patients were treated in the Intensive Care Unit (ICU) from 34 h to 45 d, while 3 were found dead by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service