Skip to Content
Merck
All Photos(2)

Documents

45569

Supelco

Methacrifos

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13O5PS
CAS Number:
Molecular Weight:
240.21
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

COC(=O)\C(C)=C\OP(=S)(OC)OC

InChI

1S/C7H13O5PS/c1-6(7(8)9-2)5-12-13(14,10-3)11-4/h5H,1-4H3/b6-5+

InChI key

NTAHCMPOMKHKEU-AATRIKPKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chung-Pei Chang et al.
Molecules (Basel, Switzerland), 25(7) (2020-03-29)
Exposure to residues of antibiotics (e.g., sulfonamides) and insecticides (e.g., organophosphorus insecticides) in aquacultured food can adversely affect humans and animals and thus affect public health globally. Here, using a validated method, we examined the levels of residues of 12
Khaled El Hawari et al.
Food additives & contaminants. Part B, Surveillance, 12(2), 81-89 (2019-01-16)
In Lebanon, apples are among the most consumed commodities; however, pesticide residues in apples have not been evaluated so far. Therefore, this study presents the occurrence of pesticide residues in apples produced in Lebanon during 2012-2016. A total of 212
Bartosz Sell et al.
Journal of analytical toxicology, 42(3), 183-191 (2017-12-02)
Reported here is a new analytical multiclass method based on QuEChERS technique, which has proven to be effective in diagnosing fatal poisoning cases in animals. This method has been developed for the determination of analytes in liver samples comprising rodenticides
Robin Guibal et al.
Journal of chromatography. A, 1387, 75-85 (2015-02-24)
The aim of this study was first to develop and validate an analytical method for the quantification of 35 polar pesticides and 9 metabolites by ultra-high-performance-liquid chromatography combined with a high resolution time-of-flight mass spectrometer detector (UHPLC-(Q)-TOF). Various analytical conditions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service