Skip to Content
Merck
All Photos(3)

Key Documents

360597

Sigma-Aldrich

2,2,4-Trimethylpentane

ACS reagent, ≥99.0%

Synonym(s):

Isooctane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2C(CH3)3
CAS Number:
Molecular Weight:
114.23
Beilstein:
1696876
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

vapor density

3.9 (vs air)

vapor pressure

41 mmHg ( 21 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

745 °F

expl. lim.

6 %

impurities

≤0.0003 meq/g Water-soluble titr. acid
≤0.005% S compounds

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.391 (lit.)

bp

98-99 °C (lit.)

mp

−107 °C (lit.)

solubility

water: insoluble

density

0.692 g/mL at 25 °C (lit.)

SMILES string

CC(C)CC(C)(C)C

InChI

1S/C8H18/c1-7(2)6-8(3,4)5/h7H,6H2,1-5H3

InChI key

NHTMVDHEPJAVLT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,2,4-Trimethylpentane (TMP, isooctane), a volatile organic solvent is used to improve the octane rating of combustion fuels. TMP is a primary reference fuel with an arbitrary octane number of 100 as it burns smoothly without resulting in any knocking. It also a component of gasoline. The physical characteristics of the binary mixtures of n-heptane and 2,2,4-trimethylpentane has been studied that helps in understanding the combustion process of mixtures of primary reference fuels.

Application

2,2,4-Trimethylpentane (TMP, isooctane) may be used in the following studies:
  • Synthesis of polyethyleneimine /silica hybrid nanoparticles.
  • Amino acid quantification process.
  • To dissolve di(2-ethylhexyl) phosphoric acid(DEHPA) to form a solution that is used in the extraction and back extraction of methylene blue.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sulforhodamine B-Loaded Polyethyleneimine/Silica Hybrid Nanoparticles.
Blumling III and Silva GA.
American Journal of Neuroprotection and Neuroregeneration, 6.1, 73-77 (2014)
Paul Hellier et al.
Frontiers in bioengineering and biotechnology, 3, 49-49 (2015-05-06)
The metabolic engineering of photosynthetic microbes for production of novel hydrocarbons presents an opportunity for development of advanced designer biofuels. These can be significantly more sustainable, throughout the production-to-consumption lifecycle, than the fossil fuels and crop-based biofuels they might replace.
Density, Viscosity, Speed of Sound, Bulk Modulus, and Surface Tension of Binary Mixtures of n-Heptane+ 2,2,4-Trimethylpentane at (293.15 to 338.15) K and 0.1 MPa.
Prak DJL, et al.
Journal of Chemical and Engineering Data, 59(11), 3842-3851 (2014)
Rafael Bermúdez et al.
PloS one, 10(5), e0123945-e0123945 (2015-05-15)
The unabated rise in anthropogenic CO₂ emissions is predicted to strongly influence the ocean's environment, increasing the mean sea-surface temperature by 4°C and causing a pH decline of 0.3 units by the year 2100. These changes are likely to affect
Reversed Micellar Extraction of Methylene Blue by using Di (2-ethylhexyl) Phosphoric Acid.
Kinugasa T, et al.
Solvent Extraction Research and Development, Japan, 22(2), 169-176 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service