Skip to Content
AG
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2,2,4-Trimethylpentane
Synonym(s):
Isooctane
Linear Formula:
(CH
3
)
2
CHCH
2
C(CH
3
)
3
CAS Number:
540-84-1
Molecular Weight:
114.23
EC Number:
208-759-1
Products
Product Category
solvents (8)
Brand
Sigma-Aldrich (9)
Supelco (4)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
ACS reagent (1)
Anhydrous (1)
Puriss (1)
Puriss P.A. (1)
analytical standard (1)
Technique
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
360597
2,2,4-Trimethylpentane,
ACS reagent, ≥99.0%
Expand
View Pricing
34862
2,2,4-Trimethylpentane,
suitable for HPLC, ≥99%
Expand
View Pricing
650439
2,2,4-Trimethylpentane,
HPLC Plus, for HPLC, GC, and residue analysis, ≥99.5%
Expand
View Pricing
32291
2,2,4-Trimethylpentane,
puriss. p.a., ≥99.5% (GC)
Expand
View Pricing
258776
2,2,4-Trimethylpentane,
ReagentPlus
®
, ≥99%
Expand
View Pricing
15-2190
2,2,4-Trimethylpentane,
JIS special grade
Expand
View Pricing
TX1389
2,2,4-Trimethylpentane,
OmniSolv
®
Expand
View Pricing
360066
2,2,4-Trimethylpentane,
anhydrous, 99.8%
Expand
View Pricing
59030
2,2,4-Trimethylpentane,
analytical standard
Expand
View Pricing
TX1395
2,2,4-Trimethylpentane,
Practical
Expand
View Pricing
TX1390
2,2,4-Trimethylpentane,
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS
Expand
View Pricing
15-2200
2,2,4-Trimethylpentane,
SAJ first grade, ≥98.0%
Expand
View Pricing
15-2210
2,2,4-Trimethylpentane,
≥99.5%, suitable for absorption spectrum analysis
Expand
View Pricing