Skip to Content
Merck
All Photos(1)

Key Documents

29210

Supelco

Eucalyptol

analytical standard

Synonym(s):

1,3,3-Trimethyl-2-oxabicyclo[2.2.2]octane, 1,8-Cineole, 1,8-Epoxy-p-menthane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Beilstein:
105109
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.457 (lit.)
n20/D 1.458

bp

176-177 °C (lit.)

mp

1-2 °C (lit.)

density

0.921 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

C[C@]12CC[C@H](CC1)C(C)(C)O2

InChI

1S/C10H18O/c1-9(2)8-4-6-10(3,11-9)7-5-8/h8H,4-7H2,1-3H3/t8-,10+

InChI key

WEEGYLXZBRQIMU-WAAGHKOSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Eucalyptol is a saturated monoterpene. It is a volatile oil with medicinal values naturally found in eucalyptus oil. It is a isoprenoid with 2-isoprene subunits (C10).[1]

Application

Eucalyptol may be used as a reference standard for the determination of the analyte in:
  • Melon fruits by headspace solid-phase microextraction (HS-SPME) and gas chromatography-mass spectrometry.[2]
  • Chrysanthemum flower by pressurized hot water extraction (PHWE) followed by headspace solid-phase microextraction (HS-SPME) and gas chromatography-mass spectrometry.[3]
  • Pharmaceutical formulations by full evaporation technique extraction followed by capillary gas chromatography and ion-trap detection.[4]
  • External analgesic pharmaceutical formulations by densitometric HPTLC analysis.[5]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
It has been used as reference standard in densitometric HPTLC analysis for the determination of Eucalyptol from external analgesic pharmaceutical formulations.[5]

Other Notes

This compound is commonly found in plants of the genus: achillea angelica cinnamomum curcuma eucalyptus mentha salvia thymus valeriana zingiber

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anti-inflammatory activity of 1.8-cineol (eucalyptol) in bronchial asthma: a double-blind placebo-controlled trial.
Juergens, U. R., et al.
Respiratory Medicine, 97.3, 250-256 (2003)
Fast quantitative determination of aroma volatile constituents in melon fruits by headspace--solid-phase microextraction and gas chromatography-mass spectrometry.
Verzera A, et al.
Food Analytical Methods, 4(2), 141-149 (2011)
Volatile organic compounds determined in pharmaceutical products by full evaporation technique and capillary gas chromatography/ion-trap detection.
Schuberth, Jan
Analytical Chemistry, 68(8), 1317-1320 (1996)
Gas chromatography-mass spectrometry following pressurized hot water extraction and solid-phase microextraction for quantification of eucalyptol, camphor, and borneol in Chrysanthemum flowers.
Dong L, et al.
Journal of Separation Science, 30(1), 86-89 (2007)
Sara Invitto et al.
Respiratory physiology & neurobiology, 259, 37-44 (2018-07-15)
Obstructive Sleep Apnea Syndrome (OSA) is characterized by snoring associated with repeated apnea and/or obstructive hypopnea. The nasal airways of OSA patients, measured via acoustic rhinometry, could be significantly narrower than healthy subjects and this reduced nasal structure can impair

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service