Skip to Content
Merck
All Photos(1)

Key Documents

51782

Supelco

Linalool

analytical standard

Synonym(s):

(±)-3,7-Dimethyl-1,6-octadien-3-ol, (±)-3,7-Dimethyl-3-hydroxy-1,6-octadiene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)(OH)CH=CH2
CAS Number:
Molecular Weight:
154.25
Beilstein:
1721488
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor pressure

0.17 mmHg ( 25 °C)

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.462 (lit.)

bp

194-197 °C/720 mmHg (lit.)

density

0.87 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

C\C(C)=C\CCC(C)(O)C=C

InChI

1S/C10H18O/c1-5-10(4,11)8-6-7-9(2)3/h5,7,11H,1,6,8H2,2-4H3

InChI key

CDOSHBSSFJOMGT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Certan Vial
Linalool is a principle monoterpene component present in many essential oils of aromatic species[1], which possess anti-inflammatory property and hence can be used to cure a variety of ailments, found to be both acute and chronic.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: cinnamomum salvia thymus

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

171.0 °F - Pensky-Martens closed cup

Flash Point(C)

77.2 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anti-inflammatory activity of linalool and linalyl acetate constituents of essential oils
Peana.T.A, et al.
Phytomedicine, 9(8), 721-726 (2002)
Linalool modifies the nicotinic receptor-ion channel kinetics at the mouse neuromuscular junction
Re.L, et al.
Pharmacological Research, 42(2), 177-181 (2000)
L Re et al.
Pharmacological research, 42(2), 177-182 (2000-07-11)
Linalool is a monoterpene compound reported to be a major component of essential oils in various aromatic species. Several linalool-producing species are used in traditional medical systems. Among these is Aeolanthus suaveolens G. Dom (Labiatae) which is used as an
C S Letizia et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 41(7), 943-964 (2003-06-14)
A toxicologic and dermatologic review of linalool when used as a fragrance ingredient, is presented.
Ali M Al Hazmi et al.
Organic letters, 16(19), 5104-5107 (2014-09-17)
The permanganate-mediated oxidative cyclization of a series of 2-methylenehept-5-eneoates bearing different chiral auxiliaries was investigated, leading to the discovery of trans-2-tritylcyclohexanol (TTC) as a highly effective chiral controller for the formation of the 2,5-substituted THF diol product with high diastereoselectivity

Articles

A throughput HS-SPME-GC-MS method to analyze volatile compounds in Perilla leaves at low extraction temperatures using four different SPME fibers.

A throughput HS-SPME-GC-MS method to analyze volatile compounds in Perilla leaves at low extraction temperatures using four different SPME fibers.

A throughput HS-SPME-GC-MS method to analyze volatile compounds in Perilla leaves at low extraction temperatures using four different SPME fibers.

A throughput HS-SPME-GC-MS method to analyze volatile compounds in Perilla leaves at low extraction temperatures using four different SPME fibers.

Protocols

Fast GC analysis of sweet orange essential oil in hexane. Key components identified includes: β-Farnesene; α-Huµlene; Germacrene D; (+)-Valencene; Bicyclogermacrene; (+)-δ-Cadinene

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service