Skip to Content
Merck
All Photos(1)

Documents

00020590

Supelco

1,8-Cineole

primary reference standard

Synonym(s):

Eucalyptol, 1,3,3-Trimethyl-2-oxabicyclo[2.2.2]octane, 1,8-Cineole, 1,8-Epoxy-p-menthane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Beilstein:
105109
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

form

liquid

CofA

certificate is enclosed in each package

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

refractive index

n20/D 1.457 (lit.)

bp

176-177 °C (lit.)

mp

1-2 °C (lit.)

density

0.921 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

C[C@]12CC[C@H](CC1)C(C)(C)O2

InChI

1S/C10H18O/c1-9(2)8-4-6-10(3,11-9)7-5-8/h8H,4-7H2,1-3H3/t8-,10+

InChI key

WEEGYLXZBRQIMU-WAAGHKOSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,8-Cineole is the main constituent of essential oils. It is a terpene oxide. It is used as percutaneous penetration enhancer, skin stimulant in skin baths, treating bronchitis, sinusitis and rheumatism. It has decongestant and antitussive property.
Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

1,8-Cineole may have been used in quantitative analysis of volatile and semi-volatile compounds of Salvia officinalis using GC-MS method.
Reference Standard in the analysis of herbal medicinal products

Other Notes

This compound is commonly found in plants of the genus: achillea angelica cinnamomum curcuma eucalyptus mentha salvia thymus valeriana zingiber

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antiinflammatory and antinociceptive effects of 1,8-cineole a terpenoid oxide present in many plant essential oils.
Santos FA, Rao VS.
Phytotherapy Research, 14(4), 240-244 (2000)
Capillary gas chromatography-mass spectrometry of volatile and semi-volatile compounds of Salvia officinalis.
Radulescu V, Chiliment S, Oprea E.
Journal of Chromatography A, 1027(1-2), 121-126 (2004)
Anke Fähnrich et al.
Plant molecular biology, 85(1-2), 135-145 (2014-02-05)
Nicotiana species of the section Alatae emit a characteristic floral scent comprising the' cineole cassette' monoterpenes 1,8-cineole, limonene, myrcene, β-pinene, α-pinene, sabinene and α-terpineol. All previously isolated 'cineole cassette'-monoterpene synthase genes are multi product enzymes that synthesize the seven compounds
Zerihun A Demissie et al.
Plant molecular biology, 79(4-5), 393-411 (2012-05-18)
Several members of the genus Lavandula produce valuable essential oils (EOs) that are primarily constituted of the low molecular weight isoprenoids, particularly monoterpenes. We isolated over 8,000 ESTs from the glandular trichomes of L. x intermedia flowers (where bulk of
Hee-jin Jun et al.
Bioorganic & medicinal chemistry letters, 23(2), 579-583 (2012-12-19)
We investigated the effect of cineole on the expression of genes related to reverse cholesterol transport and hepatic fatty acid metabolism. Cineole, a small aroma compound in teas and herbs, significantly stimulated the transactivation of liver X receptor modulator (LXR)-α

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service