Skip to Content
Merck
All Photos(1)

Documents

28125

Sigma-Aldrich

18-Crown-6

purum, ≥99.0% (GC)

Synonym(s):

18C6, 1,4,7,10,13,16-Hexaoxacyclooctadecane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H24O6
CAS Number:
Molecular Weight:
264.32
Beilstein:
1619616
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥99.0% (GC)

form

solid

mp

38-41 °C
42-45 °C (lit.)

SMILES string

O1CCOCCOCCOCCOCCOCC1

InChI

1S/C12H24O6/c1-2-14-5-6-16-9-10-18-12-11-17-8-7-15-4-3-13-1/h1-12H2

InChI key

XEZNGIUYQVAUSS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

18-Crown-6 can be used as a model for crown ethers to study the effect of acid-base properties of mixed solvent on their solution enthalpy.It is generally used as a phase transfer catalyst and a complexing agent. It specifically binds to potassium ion, ammonium or protonated alkyl amines to form complexes.
Can be useful as phase-transfer catalysts.

Other Notes

Macrocyclic polyethers with repeating (-CH2CH2O) units. The compounds are ionophoric (form stable complexes with cations).
Phase transfer catalyst, complexing-agent

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lower rim calix (4) arene ketone derivatives and their interaction with alkali metal cations. Structural and thermodynamic (solution and complexation) characterisation of the tetraphenyl ketone derivative and its sodium complex.
de Namor AFD, et al.
Physical Chemistry Chemical Physics, 3(18), 4010-4021 (2001)
Counter cation-controlled air oxidation of manganese derivatives of tetrachlorocatechol.
Ruiz R, et al.
Inorganic Chemistry Communications, 3(2), 76-79 (2000)
Separation of transition metal cations by capillary electrophoresis optimization of complexing agent concentrations (lactic acid and 18-crown-6).
Francois C, et al.
Journal of Chromatography A, 717(1-2), 393-408 (1995)
Chiral separations by host-guest complexation with cyclodextrin and crown ether in capillary zone electrophoresis.
Kuhn R, et al.
Chromatographia, 33(1-2), 32-36 (1992)
Effect of base-acid properties of the mixtures of water with methanol on the solution enthalpy of selected cyclic ethers in this mixture at 298.15 K.
Jozwiak M and Warczakowska A
Journal of Thermal Analysis and Calorimetry, 121(2), 765-770 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service