Skip to Content
Merck
All Photos(1)

Documents

31589

Sigma-Aldrich

Diethanolamine

BioUltra, ≥99.5% (GC)

Synonym(s):

2,2′-Iminodiethanol, Bis(2-hydroxyethyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HN(CH2CH2OH)2
CAS Number:
Molecular Weight:
105.14
Beilstein:
605315
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

vapor density

3.6 (vs air)

vapor pressure

<0.98 atm ( 100 °C)

product line

BioUltra

Assay

≥99.5% (GC)

form

liquid
viscous liquid

autoignition temp.

689 °F

expl. lim.

10.6 %

impurities

insoluble matter, passes filter test
≤0.2% water

refractive index

n20/D 1.477 (lit.)

pH

11.0-12.0 (25 °C, 1 M in H2O)

bp

217 °C/150 mmHg (lit.)

mp

28 °C (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

density

1.097 g/mL at 25 °C (lit.)

anion traces

carbonate (CO32-): ≤2000 mg/kg

cation traces

Al: ≤1 mg/kg
As: ≤0.1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Li: ≤1 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.04
λ: 280 nm Amax: 0.02

SMILES string

OCCNCCO

InChI

1S/C4H11NO2/c6-3-1-5-2-4-7/h5-7H,1-4H2

InChI key

ZBCBWPMODOFKDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Component in diethanolamine-hydrochloric acid buffer (pH 7.80 - 9.90); Improved chemiluminescent detection of alkaline phosphatase.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Oral

Target Organs

Kidney,Liver,Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved chemiluminescent detection of alkaline phosphatase.
I Bronstein et al.
BioTechniques, 9(2), 160-161 (1990-08-01)
D.D. Perrin et al.
Buffers for pH and Metal Ion Control, 145-145 (1974)
William Conway et al.
Environmental science & technology, 46(13), 7422-7429 (2012-05-25)
The kinetics of the fast reversible carbamate formation reaction of CO(2)(aq) with a series of substituted cyclic secondary amines as well as the noncyclic secondary amine diethanolamine (DEA) has been investigated using the stopped-flow spectrophotometric technique at 25.0 °C. The
Siou-Yin Chen et al.
Inorganic chemistry, 51(8), 4448-4457 (2012-04-05)
The synthesis, X-ray crystallography, magnetic properties, and high-field electron paramagnetic resonance (HFEPR) of a new heptanuclear manganese complex [Mn(7)(heamp)(6)](ClO(4))(2)·4CH(2)Cl(2)·H(2)O (complex 2), in which heampH(3) is 2-[N,N-di(2-hydroxyethyl)aminomethyl]phenol (compound 1), is reported. Complex 2 has a hexagonal, disk-shaped topology and contains six
Honghui Yu et al.
Journal of immunology (Baltimore, Md. : 1950), 193(6), 2902-2910 (2014-08-15)
Anti-D can prevent immunization to the RhD Ag on RBCs, a phenomenon commonly termed Ab-mediated immune suppression (AMIS). The most accepted theory to explain this effect has been the rapid clearance of RBCs. In mouse models using SRBC, these xenogeneic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service