Skip to Content
Merck
All Photos(4)

Documents

194123

Sigma-Aldrich

Benzimidazole

98%

Synonym(s):

1,3-Benzodiazole, BZI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6N2
CAS Number:
Molecular Weight:
118.14
Beilstein:
109682
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
32151902
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

169-171 °C (lit.)

solubility

xylene: soluble 1g in 2g (boiling)(lit.)
alcohol: freely soluble(lit.)
benzene: insoluble(lit.)
cold water: very slightly soluble(lit.)
diethyl ether: very slightly soluble(lit.)
petroleum ether: insoluble(lit.)
water: soluble (hot)(lit.)

SMILES string

c1ccc2[nH]cnc2c1

InChI

1S/C7H6N2/c1-2-4-7-6(3-1)8-5-9-7/h1-5H,(H,8,9)

InChI key

HYZJCKYKOHLVJF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Benzimidazole was used as template compound in the preparation of molecularly imprinted polymer via electropolymerization and electrodeposition of pyrrole on a pencil graphite electrode. It was also used in the preparation of benzimidazolium surfactant, 1-hexadecyl-1H-benzimidazole.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Christina Mark et al.
Nuclear medicine and biology, 40(2), 295-303 (2012-12-26)
The norepinephrine transporter (NET) is an important target for research in neurology and psychology and is involved in the pathophysiology of many neurodegenerative diseases such as Alzheimer's disease and attention deficient hyperactivity disorder. For visualization of NET abundance and deregulation
Nevine M Y Elsayed et al.
Bioorganic chemistry, 82, 340-359 (2018-11-15)
VEGFR-2 has a pivotal role in promoting cancer angiogenesis. Herein, two series of novel indazole-based derivatives were designed, synthesized and evaluated for their in vitro inhibitory action against VEGFR-2 kinase enzyme. The second series 11a-e exhibited better potency than the
Stephan Mokesch et al.
Molecules (Basel, Switzerland), 24(13) (2019-06-30)
Fine-tuning of the properties of a recently reported 1,3-indandione-based organoruthenium complex is attempted to optimize the stability under physiological conditions. Previous work has shown its capacity of inhibiting topoisomerase IIα; however, fast aquation leads to undesired reactions and ligand cleavage
Benzimidazolium surfactants for modification of clays for use with styrenic polymers.
Costache MC, et al.
Polymer Degradation and Stability, 92(10), 1753-1762 (2007)
Wang Zhang et al.
ACS nano, 14(2), 1971-1981 (2020-01-31)
The development of highly efficient electrocatalysts to reduce overpotentials is vital for accelerating the sluggish oxygen evolution reaction (OER) processes. Herein, we demonstrate ultrathin heterogeneous nanosheets as a promising OER electrocatalyst, which are composed of ultrafine CoFeOx nanoparticles and a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service