177806
Tetraethylammonium hydroxide solution
20 wt. % in H2O
Synonym(s):
TEA hydroxide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(C2H5)4N(OH)
CAS Number:
Molecular Weight:
147.26
Beilstein:
3912961
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Recommended Products
form
liquid
Quality Level
concentration
20 wt. % in H2O
refractive index
n20/D 1.373
density
1.01 g/mL at 25 °C
functional group
amine
SMILES string
[OH-].CC[N+](CC)(CC)CC
InChI
1S/C8H20N.H2O/c1-5-9(6-2,7-3)8-4;/h5-8H2,1-4H3;1H2/q+1;/p-1
InChI key
LRGJRHZIDJQFCL-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
General description
The product is 20wt.% solution of tetraethylammonium hydroxide (TEA hydroxide, TENOH) in H2O. TEA hydroxide is a quaternary ammonium hydroxide (QAH). It participates in the synthesis of zeolite beta. TENOH peptized titania aqueous sols have been employed in the preparation of nanocrystalline anatase films.
Application
- Chromatographic applications: It plays a role in the preparation of ferrofluid from adipic acid-coated magnetic nanoparticles and hydrophobic deep eutectic solvent used in dispersive micro-solid-phase extraction, enhancing the pre-concentration and determination of clozapine in biological samples (Nia and Hadjmohammadi, 2023).
- Material and interfacial science: Utilized in the remote control of polyelectrolyte-surfactant mixtures, tetraethylammonium hydroxide solution aids in creating responsive materials and interfacial properties for innovative material science applications (Schnurbus et al., 2022).
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Crystallization mechanism of zeolite beta from (TEA)2O, Na2O and K2O containing aluminosilicate gels.
Perez-Pariente J, et al.
Applied Catalysis, 31(1), 35-64 (1987)
Izumi C Mori et al.
Chemosphere, 120, 299-304 (2014-08-26)
The aquatic ecotoxicity of chemicals involved in the manufacturing process of thin film transistor liquid crystal displays was assessed with a battery of four selected acute toxicity bioassays. We focused on tetramethylammonium hydroxide (TMAH, CAS No. 75-59-2), a widely utilized
Hydrothermal processing of nanocrystalline anatase films from tetraethylammonium hydroxide peptized titania sols.
Yang J, et al.
J. Eur. Ceram. Soc., 24(2), 335-339 (2004)
Nanomechanics of cation-π interactions in aqueous solution.
Qingye Lu et al.
Angewandte Chemie (International ed. in English), 52(14), 3944-3948 (2013-03-02)
Dong-Mei Zhang et al.
PloS one, 7(10), e47030-e47030 (2012-10-12)
Cajaninstilbene acid (CSA) is a major active component present in the leaves of Cajanus cajan (L.) Millsp. The present study explores the underlying cellular mechanisms for CSA-induced relaxation in rat renal arteries. Vascular reactivity was examined in arterial rings that
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service