Skip to Content
Merck
All Photos(3)

Key Documents

227048

Sigma-Aldrich

Cyclohexane

anhydrous, 99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12
CAS Number:
Molecular Weight:
84.16
Beilstein:
1900225
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
Assay:
99.5%
bp:
80.7 °C (lit.)
vapor pressure:
168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)

grade

anhydrous

Quality Level

vapor density

2.9 (vs air)

vapor pressure

168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)

Assay

99.5%

form

liquid

autoignition temp.

500 °F

expl. lim.

9 %

impurities

<0.001% water
<0.005% water (100 mL pkg)

evapn. residue

<0.0005%

color

colorless

refractive index

n20/D 1.426 (lit.)

bp

80.7 °C (lit.)

mp

4-7 °C (lit.)

density

0.779 g/mL at 25 °C (lit.)

SMILES string

C1CCCCC1

InChI

1S/C6H12/c1-2-4-6-5-3-1/h1-6H2

InChI key

XDTMQSROBMDMFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyclohexane is a cyclic alkane that predominantly exists in chair conformation due to higher stability.

Application

Cyclohexane may undergo oxidation using hydrogen peroxide as oxidant and in the presence of a polyoxotungstate catalyst to form cyclohexanone and cyclohexanol as the main products. Its dehydrogenation in the presence of Ni-Cu/SiO2 catalysts forms benzene in good yield.

Other Notes

The article number 227048-4X2L will be discontinued. Please order the single bottle 227048-2L which is physically identical with the same exact specifications.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tina Gulin-Sarfraz et al.
Pharmaceutics, 11(4) (2019-04-04)
Diseases in the respiratory tract rank among the leading causes of death in the world, and thus novel and optimized treatments are needed. The lungs offer a large surface for drug absorption, and the inhalation of aerosolized drugs are a
Oxidation of cyclohexane and ethylbenzene by hydrogen peroxide over Co-substituted heteropolytungstate catalyst.
Kanjina W and Trakarnpruk W
3 Biotech, 20(2) (2017)
Nataša Šegatin et al.
Foods (Basel, Switzerland), 9(7) (2020-07-12)
The dielectric properties of six refined edible oils with different fatty-acid compositions were determined for oils incubated at 180 °C up to 40 h. The oil degradation was evaluated by the dielectric dispersion and dielectric loss in the frequency range
Cyclohexane dehydrogenation over Ni-Cu/SiO 2 catalyst: Effect of copper addition.
Xia Z, et al.
Catalysis Communications, 90, 39-42 (2017)
Samuel Kim et al.
Methods in enzymology, 472, 119-132 (2010-06-29)
Microfluidics serves as a convenient platform for single-molecule experiments by providing manipulation of small amounts of liquids and micron-sized particles. An adapted version of capillary electrophoresis (CE) on a microchip can be utilized to separate chemical species with high resolution

Articles

Substances are said to be miscible in one another if they dissolve to form a uniform solution. Bookmark or download our miscibility table for common lab solvents.

Substances are said to be miscible in one another if they dissolve to form a uniform solution. Bookmark or download our miscibility table for common lab solvents.

Substances are said to be miscible in one another if they dissolve to form a uniform solution. Bookmark or download our miscibility table for common lab solvents.

Substances are said to be miscible in one another if they dissolve to form a uniform solution. Bookmark or download our miscibility table for common lab solvents.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service