Skip to Content
Merck
All Photos(2)

Key Documents

Z568023

PureSolv Micro 100 Liter solvent purification system

with activated Alumina column, joint: ST/NS 24/40 US

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.85
Pricing and availability is not currently available.

material

anodized aluminum frame
stainless steel fittings
stainless steel tubing

Joint

joint ST/NS29/32 Stainless Steel
joint: ST/NS24/40 US

description

Column valve: Swagelok Stainless Steel, 3-way
Leak Rate- no detectable leaks with helium mass spectrometer (sensitivity of 5 x 10-6 std cc/sec)
Scrubbing capacity- 5 weight % water absorption, approx 100 L solvent

manufacturer/tradename

Innovative Technologies

parameter

5 psi pressure

Looking for similar products? Visit Product Comparison Guide

General description

"Easy access to smaller volumes of anhydrous solvent"
The Pure Solv Micro solvent purification system is designed as both an economic and space friendly alternative to the larger more traditional free standing units, only 12 in. W x 13 in. D x 19 in. H. This purification system will remove water from approximately 100 L of solvent using the same proven design as the Innovative Technology, Inc. Pure Solv MD system. Pure Solv Micro is perfect for the researcher with modest requirements for a single anhydrous solvent who prefers the convenience of having a dedicated solvent drying and dispensing station. No longer do you have to share; now you can have your own.

  • Pure Solv Micro has been packaged so that it can be placed on a bench top, inside a fume hood or flammable cabinet as desired by the researcher in charge
  • Pure Solv Micro facilitates proper air free dispensing techniques using an integrated gas and vacuum manifold system. A KNF Laboport solid PTFE vacuum pump, 20 L pumping speed or house vacuum is recommended for use with Pure Solv systems
  • All systems include a size 24/40 or 29/32 stainless steel glassware adapter
  • Small capacity 4 L reservoirs include custom designed, leak-tight, clamp style cap and Swagelok quick-disconnect valves for easy handling and refilling
  • Pure Solv Micro is bonded and grounded to remove hazards associated with electrostatic discharge and ensure safe operating environment
  • Exhaust connections available
  • Inert gases used: Nitrogen- Argon
  • Purifying material: Based on solvent type-order separately
  • Attainable purity: Low PPM H2O and O2
  • Solvent volume per charge on the column: approx. 50 mL
  • Solvent residency time on the column: approx. 4 h


ORDERING NOTE: Order Pure Solv Micro with the appropriate activated purification column. Solvent purification columns are disposable. Additional activated columns may be ordered when replacement is necessary. Order spill tray separately.

Linkage

Use the PureSolv Selection Guide to determine which system is needed. Go to the PureSolv Overview to see details of the system.

Legal Information

Pure Solv is a trademark of Inert Corp.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service